Skip to Content
Merck
All Photos(1)

Key Documents

50892

Sigma-Aldrich

Octanoyl-L-carnitine

≥97.0% (TLC)

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxooctyl)oxy]-1-propanaminium inner salt, L-Carnitine octanoyl ester, C8-Carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29NO4
CAS Number:
Molecular Weight:
287.40
Beilstein:
5955761
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (TLC)

optical activity

[α]/D -17±2°, c = 0.1 in H2O

impurities

≤10% water

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCC)=O)CC([O-])=O

InChI

1S/C15H29NO4/c1-5-6-7-8-9-10-15(19)20-13(11-14(17)18)12-16(2,3)4/h13H,5-12H2,1-4H3/t13-/m1/s1

InChI key

CXTATJFJDMJMIY-CYBMUJFWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Octanoylcarnitine is the physiologically active form of octanoylcarnitine and is detected in medium-chain acyl-CoA dehydrogenase (MCAD) deficiency. MCAD is characterized by intolerance to prolonged fasting, recurrent episodes of hypoglycemic coma with medium-chain dicarboxylic aciduria, impaired ketogenesis, and low plasma and tissue carnitine levels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David S Millington et al.
Methods in molecular biology (Clifton, N.J.), 708, 55-72 (2011-01-06)
The acylcarnitine profile is a diagnostic test for inherited disorders of fatty acid and branched-chain amino acid catabolism. Patients with this type of metabolic disorder accumulate disease-specific acylcarnitines that correlate with the acyl coenzyme A compounds in the affected mitochondrial
J Will Thompson et al.
Rapid communications in mass spectrometry : RCM, 26(21), 2548-2554 (2012-09-26)
Acylcarnitines are routinely analyzed by electrospray ionization tandem mass spectrometry (ESI-MS/MS) both in clinical diagnostic and public health newborn screening laboratories from plasma and dried whole blood spots (DBS) on filter paper. The use of DBS as a convenient method
Steen Larsen et al.
The Journal of physiology, 590(14), 3349-3360 (2012-05-16)
Skeletal muscle mitochondrial content varies extensively between human subjects. Biochemical measures of mitochondrial proteins, enzyme activities and lipids are often used as markers of mitochondrial content and muscle oxidative capacity (OXPHOS). The purpose of this study was to determine how
Josiane Joseph et al.
Metabolites, 8(4) (2018-10-05)
Duchenne muscular dystrophy (DMD) is a musculoskeletal disorder that causes severe morbidity and reduced lifespan. Individuals with DMD have an X-linked mutation that impairs their ability to produce functional dystrophin protein in muscle. No cure exists for this disease and
Judit Bene et al.
World journal of gastroenterology, 11(42), 6671-6675 (2006-01-21)
To determine the fasting plasma carnitine ester in patients with celiac disease. We determined the fasting plasma carnitine ester profile using ESI triple quadrupol mass spectrometry in 33 adult patients with biopsy-confirmed maturity onset celiac disease maintained on long term

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service