Skip to Content
Merck
All Photos(2)

Documents

36772

Supelco

Fenpropimorph

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H33NO
CAS Number:
Molecular Weight:
303.48
Beilstein:
8164441
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

storage condition

under inert gas

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(CN1CC(C)OC(C)C1)Cc2ccc(cc2)C(C)(C)C

InChI

1S/C20H33NO/c1-15(12-21-13-16(2)22-17(3)14-21)11-18-7-9-19(10-8-18)20(4,5)6/h7-10,15-17H,11-14H2,1-6H3

InChI key

RYAUSSKQMZRMAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>212.0 °F - closed cup

Flash Point(C)

> 100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Young Park et al.
The plant pathology journal, 36(6), 579-590 (2020-12-15)
Botrytis cinerea, which causes gray mold disease in more than 200 plant species, is an economically important pathogen that is mainly controlled by synthetic fungicides. Synergistic fungicide mixtures can help reduce fungicide residues in the environment and mitigate the development
Amandine Bordat et al.
BMC plant biology, 17(1), 167-167 (2017-10-21)
Phoma macdonaldii has been reported as the causal agent of black stem disease (BS) and premature ripening (PR) on sunflower. PR is considered as the most widespread and detrimental disease on sunflower in France. While genetic variability and QTL mapping
Loreen Sommermann et al.
PloS one, 13(4), e0195345-e0195345 (2018-04-06)
Fungal communities in agricultural soils are assumed to be affected by soil and crop management. Our intention was to investigate the impact of different tillage and fertilization practices on fungal communities in a long-term crop rotation field trial established in
Daniel Degreif et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1864(7), 1005-1016 (2019-03-29)
Cells maintain physicochemical characteristics of membranes in order to allow for proper function of membrane-associated cellular processes, such as endocytosis and exocytosis. To investigate the interplay between membrane properties and biological processes, we applied lipid engineering approaches that allowed for
Xuegui Wang et al.
Frontiers in microbiology, 9, 2591-2591 (2018-11-15)
The pathogen Botrytis cinerea is a very dangerous pathogen that infects many economically important crops such as grape, strawberry, tomato, and eggplant. Cyprodinil, a pyrimidine amine fungicide, and fenhexamid, an amide fungicide, are new reagents for controlling gray mold with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service