Skip to Content
Merck
All Photos(1)

Documents

32013

Supelco

Tebuconazole

PESTANAL®, analytical standard

Synonym(s):

1-(4-Chlorophenyl)-4,4-dimethyl-3-(1H-1,2,4-triazol-1-ylmethyl)-3-pentanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22ClN3O
CAS Number:
Molecular Weight:
307.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)(C)C(CO)(CCc1ccc(Cl)cc1)n2cncn2

InChI

1S/C16H22ClN3O/c1-15(2,3)16(10-21,20-12-18-11-19-20)9-8-13-4-6-14(17)7-5-13/h4-7,11-12,21H,8-10H2,1-3H3

InChI key

UAJBSHCDOPGGBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tebuconazole is a triazole fungicide. It is mostly used to control diseases in agricultural crops and is effective against diseases which are soil borne and foliar fungal based.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tebuconazole may have been used as reference standard in gas chromatography–mass spectrometry in selected ion monitoring mode (GC/MS, SIM) method for determination of pesticides from Hyptis pectinata, medicinal plant using [Zn(BDC)(H2O)2]n as adsorbent.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coordination polymer adsorbent for matrix solid-phase dispersion extraction of pesticides during analysis of dehydrated Hyptis pectinata medicinal plant by GC/MS.
Aquino A et.al.
Talanta, 83(2), 631-636 (2010)
Andres Mäe et al.
Frontiers in plant science, 11, 385-385 (2020-05-01)
Septoria tritici blotch (STB) is caused by the ascomycete Zymoseptoria tritici and one of the predominating diseases in wheat (Triticum aestivum) in Europe. The control of STB is highly reliant on frequent fungicide applications. The primary objective of this study
Tebuconazole application decreases soil microbial biomass and activity.
Mu?oz-Leoz, Borja, et al.
Soil Biology and Biochemistry, 43.10, 2176-2183 (2011)
Stefanie Gerbig et al.
Analytical and bioanalytical chemistry, 409(8), 2107-2117 (2016-12-31)
Direct analysis of fruit and vegetable surfaces is an important tool for in situ detection of food contaminants such as pesticides. We tested three different ways to prepare samples for the qualitative desorption electrospray ionization mass spectrometry (DESI-MS) analysis of
Belén Patiño et al.
International journal of food microbiology, 283, 45-51 (2018-08-14)
Filamentous fungi are an invaluable source for biocontrol strategies and for production and development of different antifungal polypeptides. Within this context, cysteine-rich antifungal AFP-like peptides stand out among many different antimicrobial compounds given their production easiness, stability, versatility, and efficacy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service