Skip to Content
Merck
All Photos(1)

Key Documents

37902

Supelco

Hexaflumuron

PESTANAL®, analytical standard

Synonym(s):

N-[3,5-Dichloro-4-(1,1,2,2-tetrafluoroethoxy)phenylcarbamoyl]-2,6-difluorobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8Cl2F6N2O3
CAS Number:
Molecular Weight:
461.14
Beilstein:
8168725
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

FC(F)C(F)(F)Oc1c(Cl)cc(NC(=O)NC(=O)c2c(F)cccc2F)cc1Cl

InChI

1S/C16H8Cl2F6N2O3/c17-7-4-6(5-8(18)12(7)29-16(23,24)14(21)22)25-15(28)26-13(27)11-9(19)2-1-3-10(11)20/h1-5,14H,(H2,25,26,27,28)

InChI key

RGNPBRKPHBKNKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Huang et al.
Journal of agricultural and food chemistry, 58(8), 4839-4843 (2010-03-30)
The dissipation and residue behavior of hexaflumuron in Chinese cabbage ( Brassica pekinensis ) under different treatments were investigated at two main cabbage-growing areas in China. The dissipation rates of hexaflumuron in cabbages and soils fit a first-order decay process
J J Sheets et al.
Journal of economic entomology, 93(3), 871-877 (2000-07-21)
The rates of uptake, clearance, insect-to-insect transfer, and metabolism of [14C] hexaflumuron [N-(((3,5-dichloro-4-(1,1,2,2-tetrafluroethoxy)phenyl)- amino)carbonyl)-2,6-diflurobenzamide] were measured in eastern subterranean termite workers, Reticulitermes flavipes (Kollar), fed cellulose diets containing either 0.1 or 0.5% (wt:wt) hexaflumuron. The rate of uptake, level of
A S Sajap et al.
Journal of economic entomology, 93(2), 429-433 (2000-05-29)
Cellulosic bait matrices containing 0.5% hexaflumuron were tested against field colonies of the subterranean termite Coptotermes curvignathus Holmgren in Malaysia. Foraging activities of the termites were first monitored for several months by using survey stakes and wood bundles placed in
Nan-Yao Su
Journal of economic entomology, 100(2), 475-482 (2007-04-28)
Cellulose baits containing 0.5% hexaflumuron were hermetically sealed in a closed cell polyethylene sheet envelope and placed in soil to test their durability and efficacy against field colonies of the Formosan subterranean termite, Coptotermes formosanus Shiraki, or the eastern subterranean
N Y Su et al.
Journal of economic entomology, 93(2), 415-421 (2000-05-29)
A sensor consisting of a wooden monitor painted with a conductive circuit of silver particle emulsion was placed in a monitoring station to detect feeding activity of the subterranean termite Coptotermes havilandi Holmgren. Sensor accuracy was 100% 1 mo after

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service