Skip to Content
Merck
All Photos(1)

Key Documents

46113

Supelco

Fluazuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Cl2F5N3O3
CAS Number:
Molecular Weight:
506.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1cccc(F)c1C(=O)NC(=O)Nc2ccc(Cl)c(Oc3ncc(cc3Cl)C(F)(F)F)c2

InChI

1S/C20H10Cl2F5N3O3/c21-11-5-4-10(29-19(32)30-17(31)16-13(23)2-1-3-14(16)24)7-15(11)33-18-12(22)6-9(8-28-18)20(25,26)27/h1-8H,(H2,29,30,31,32)

InChI key

YOWNVPAUWYHLQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fluazuron insecticide research: A study on enzootic stability of tick fever in calves explored the use of fluazuron combined with fluralaner, highlighting its effectiveness in strategic tick control in tropical regions, contributing significantly to veterinary parasitology and entomology research (Zapa et al., 2024).

  • Benzoylphenyl urea Fluazuron: Research assessing the anthelmintic resistance and acaricidal efficacy of fluazuron on sheep and goat ticks demonstrated its role in managing gastrointestinal nematode resistance, supporting its use in comprehensive pest management strategies (Emsley et al., 2023).

  • Fluazuron acaricide studies: An economic evaluation of tick control strategies using fluazuron underscored its cost-effectiveness and practical application in managing drug-resistant strains of Rhipicephalus microplus, reinforcing fluazuron′s critical role in sustainable agricultural practices (Junior et al., 2022).

  • Fluazuron veterinary parasitology: An in-depth review on acaricides resistance in ticks mentioned fluazuron as a key compound in the ongoing fight against tick-borne diseases, providing insights into the mechanisms of action and resistance patterns, crucial for developing new control measures (Obaid et al., 2022).

  • Fluazuron pharmacokinetics research: A field study on rotational and selective protocols using fluazuron to control a multi-resistant strain of ticks detailed its pharmacokinetics and the subsequent impact on tick populations in Southern Brazil, offering valuable data for pharmacological and ecological studies (Centenaro et al., 2022).

Caution

Sensitive to light, handle under argon.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Welber Daniel Zanetti Lopes et al.
Parasitology international, 66(5), 627-634 (2017-05-22)
The present work consisted of eight studies to evaluate the ectoparasiticidal spectrum and determine the pharmacokinetic parameters of a pour-on combination of fipronil 1.25mg/kg+fluazuron 2.5mg/kg for cattle against Rhipicephalus microplus, Haematobia irritans and the larvae of Dermatobia hominis and Cochliomyia
U Kryger et al.
Veterinary parasitology, 143(3-4), 380-384 (2006-09-19)
While resistance against many other classes of acaricides has been described, products containing benzoylphenyl urea are currently still successfully used against the pesticide-resistant blue tick (Boophilus decoloratus) in South Africa. In order to assess any adverse impact of these tickicides
Patrícia Rosa de Oliveira et al.
Parasitology research, 112(1), 45-58 (2012-09-21)
The present study demonstrated the effects of the arthropod growth regulator, fluazuron (Acatak®), in the formation of the integument and digestive processes of Rhipicephalus sanguineus nymphs fed on rabbits treated with different doses of this chemical acaricide. For this, three
Fabrício Nascimento Gaudêncio et al.
Parasitology international, 66(5), 584-587 (2017-04-25)
The objective of this study was to assess the effect of the exposure to fluazuron on the activity of common pesticide detoxification enzyme groups in the cattle tick (Rhipicephalus microplus). Engorged females of a susceptible strain (POA) and a resistant
Dina María Beltrán Zapa et al.
Preventive veterinary medicine, 177, 104950-104950 (2020-03-24)
Rhipicephalus (Boophilus) microplus is the most important livestock tick, causing economical losses especially in tropical and subtropical regions. Pour-on formulations using synthetic chemicals, remain the most farmer friendly conventional form of tick control method. Misuse of acaricides can lead to

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service