Skip to Content
Merck
All Photos(1)

Key Documents

27655

Sigma-Aldrich

Colistin sodium methanesulfonate

from Bacillus colistinus

Synonym(s):

Colimycin sodium methanesulfonate, Methanesulfonic acid derivative of Polymyxin E, Polymyxin E sodium methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H115N16Na5O28S5
CAS Number:
Molecular Weight:
1759.90
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

biological source

Bacillus colistinus

form

powder or crystals

ign. residue

~20%

loss

≤5.0% loss on drying

color

white to off-white

pH

6.5-8.5

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CCC(C)CCCCC(=O)N[C@@H](CCN)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CCN)C(=O)N[C@H]1CCNC(=O)[C@@H](NC(=O)[C@H](CCN)NC(=O)[C@@H](CCN)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](CCN)NC1=O)C(C)O

InChI

1S/C53H100N16O13.5CH4O3S.5Na/c1-9-30(6)12-10-11-13-41(72)60-33(14-20-54)48(77)69-43(32(8)71)53(82)65-36(17-23-57)45(74)64-38-19-25-59-52(81)42(31(7)70)68-49(78)37(18-24-58)62-44(73)34(15-21-55)63-50(79)39(26-28(2)3)67-51(80)40(27-29(4)5)66-46(75)35(16-22-56)61-47(38)76;5*1-5(2,3)4;;;;;/h28-40,42-43,70-71H,9-27,54-58H2,1-8H3,(H,59,81)(H,60,72)(H,61,76)(H,62,73)(H,63,79)(H,64,74)(H,65,82)(H,66,75)(H,67,80)(H,68,78)(H,69,77);5*1H3,(H,2,3,4);;;;;/q;;;;;;5*+1/p-5/t30?,31-,32-,33+,34-,35+,36+,37+,38+,39+,40-,42+,43+;;;;;;;;;;/m1........../s1

InChI key

WSDSONZFNWDYGZ-BKFDUBCBSA-I

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Colistin sodium methanesulfate is used to permeabilize bacterial cell membranes and to study mannose-resistant haemagglutination.

Biochem/physiol Actions

Colistin sodium methanesulfate is a cyclopolypeptide antibiotic with endotoxin-binding capacity Colistin′s activity stems from the hydrophobic and hydrophillic regions. They solubilze the cytoplasmic membrane which contributes to the bactericidal effect .
Mode of Action: Binds to lipids on the cell cytoplasmic membrane of Gram-negative bacteria and disrupts the cell wall integrity.
Antimicrobial spectrum: Gram-negative bacteria.

Packaging

1g, 5g

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place. Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.J. Rogers et al.
Infection (Munich), 14, 79-79 (1986)
Mairi Ziaka et al.
Antimicrobial agents and chemotherapy, 57(4), 1938-1940 (2013-01-22)
Colistin pharmacokinetics were prospectively studied after intravenous administration of colistin methanesulphonate in critically ill patients without central nervous system infection (controls, n = 5) and in patients with external ventricular drain-associated ventriculitis after intravenous administration (EVDViv, n = 3) or
Anne H de Boer et al.
The Journal of pharmacy and pharmacology, 64(9), 1316-1325 (2012-08-14)
To use computational fluid dynamics (CFD) for evaluating and understanding the performance of the high-dose disposable Twincer™ dry powder inhaler, as well as to learn the effect of design modifications on dose entrainment, powder dispersion and retention behaviour. Comparison of
Phillip J Bergen et al.
Current opinion in pharmacology, 11(5), 464-469 (2011-08-13)
The increasing prevalence of multidrug-resistant Gram-negative bacteria worldwide has led to a re-evaluation of the previously discarded antibiotic, colistin. Despite its important role as salvage therapy for otherwise untreatable infections, dosage guidelines for the prodrug colistin methanesulfonate (CMS) are not
Stephanie J Wallace et al.
Journal of pharmaceutical sciences, 101(9), 3347-3359 (2012-05-25)
Interaction of colistin and colistin methanesulfonate (CMS) with liposomes has been studied with the view to understanding the limitations to the use of liposomes as a more effective delivery system for pulmonary inhalation of this important class of antibiotic. Thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service