Skip to Content
Merck
All Photos(2)

Key Documents

V1130

Sigma-Aldrich

Vancomycin hydrochloride from Streptomyces orientalis

BioReagent, suitable for plant cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · HCl
CAS Number:
Molecular Weight:
1485.71
Beilstein:
3704657
UNSPSC Code:
10171502
NACRES:
NA.72

biological source

Streptomyces orientalis

product line

BioReagent

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

concentration

≥900 μg/mg (as vancomycin base)

technique(s)

cell culture | plant: suitable

color

, off-white to brown or White to orange-brown

solubility

H2O: 50 mg/mL, clear, colorless to dark yellow (and Very Faint Orange to Faint Orange)

antibiotic activity spectrum

Gram-positive bacteria

application(s)

agriculture

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl[H].CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]1O[C@H]2C[C@](C)(N)[C@@H](O)[C@@H](C)O2.CN[C@H](CC(C)C)C(=O)NC3[C@H](O)c4ccc(Oc5cc6Oc7ccc(cc7Cl)[C@@H](O)[C@H]8NC(=O)[C@H](NC(=O)[C@H](NC(=O)[C@H](CC(N)=O)NC3=O)c(c5)c6)c9ccc(O)c(c9)-c%10c(O)cc(O)cc%10[C@@H](NC8=O)C(O)=O)c(Cl)c4

InChI

1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)45(31)30-13-25(6-9-36(30)80)46(60(90)77-50)74-61(91)47(28)73-59(89)35(20-43(69)82)72-62(49)92;/h6-11,13-19,23-24,34-35,42,44,46-54,56-57,65,71,78-81,83-87H,12,20-22,70H2,1-5H3,(H2,69,82)(H,72,92)(H,73,89)(H,74,91)(H,75,93)(H,76,88)(H,77,90)(H,94,95);1H/t24-,34+,35-,42+,44-,46+,47+,48-,49+,50-,51+,52+,53+,54-,56+,57+,65-,66-;/m0./s1

InChI key

LCTORFDMHNKUSG-XTTLPDOESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: glycopeptide
Vancomycin is a heptapeptide.

Application

Vancomycin hydrochloride from Streptomyces orientalis has been used as:
  • an antibiotic in screening: minimal inhibitory concentrations (MICs) Streptomyces coelicolor
  • in treating Leishmania infected sand flies
  • in combined antibiotic treatment in asthma induced mice

Biochem/physiol Actions

Mode of action: Inhibits incorporation of terminal D-alanyl-D-alanine moieties of the NAM/NAG-peptides.

Antimicrobial Spectrum: Active against Gram-positive bacteria
Vancomycin is used for treating methicillin-resistant Staphylococcus aureus (MRSA) infections and shunt-associated Staphylococcal ventriculitis. It may lead to hypersensitivity reactions like anaphylaxis and red man syndrome. Vanomycin is also used to treat thrombocytopenia and induces drug reaction with eosinophilia and systemic symptoms (DRESS) on usage.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Asano et al.
Phytopathology, 89(4), 314-319 (2008-10-24)
ABSTRACT Resting spores of Plasmodiophora brassicae were surface-disinfested by treatment with 2% chloramine-T for 20 min and then with an antibiotic solution (1,000 ppm of colistin sulfate, 1,000 ppm of vancomycin hydrochloride, and 6,000 ppm of cefotaxime sodium) for 1
Red man syndrome
Sivagnanam S and Deleu D
Critical Care, 7(2), 119-119 (2002)
Early-life antibiotics attenuate regulatory T cell generation and increase the severity of murine house dust mite-induced asthma
Adami AJ, et al.
Pediatric Research, 84(3), 426-426 (2018)
L Burgos et al.
Plant cell reports, 21(12), 1167-1174 (2003-06-06)
An improved method for adventitious regeneration from apricot leaves is described. The use of the ethylene inhibitors silver thiosulphate (30-60 micro M) or aminoethoxyvinylglycine (0.5 micro M) increased regeneration percentages in Helena and Canino apricot cultivars and also the consistency
Optimization of fermentation conditions for production of glycopeptide antibiotic vancomycin by Amycolatopsis orientalis
Padma PN, et al.
Applied Biochemistry and Biotechnology, 102(1-6), 395-405 (2002)

Articles

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service