Skip to Content
Merck
All Photos(1)

Key Documents

C1511

Sigma-Aldrich

Colistin sodium methanesulfonate

~11,500 U/mg

Synonym(s):

Colimycin sodium methanesulfonate, Methanesulfonic acid derivative of Polymyxin E, Polymyxin E sodium methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H115N16Na5O28S5
CAS Number:
Molecular Weight:
1759.90
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder

specific activity

~11,500 U/mg

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CCC(C)CCCCC(=O)N[C@@H](CCN)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CCN)C(=O)N[C@H]1CCNC(=O)[C@@H](NC(=O)[C@H](CCN)NC(=O)[C@@H](CCN)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](CCN)NC1=O)C(C)O

InChI

1S/C53H100N16O13.5CH4O3S.5Na/c1-9-30(6)12-10-11-13-41(72)60-33(14-20-54)48(77)69-43(32(8)71)53(82)65-36(17-23-57)45(74)64-38-19-25-59-52(81)42(31(7)70)68-49(78)37(18-24-58)62-44(73)34(15-21-55)63-50(79)39(26-28(2)3)67-51(80)40(27-29(4)5)66-46(75)35(16-22-56)61-47(38)76;5*1-5(2,3)4;;;;;/h28-40,42-43,70-71H,9-27,54-58H2,1-8H3,(H,59,81)(H,60,72)(H,61,76)(H,62,73)(H,63,79)(H,64,74)(H,65,82)(H,66,75)(H,67,80)(H,68,78)(H,69,77);5*1H3,(H,2,3,4);;;;;/q;;;;;;5*+1/p-5/t30?,31-,32-,33+,34-,35+,36+,37+,38+,39+,40-,42+,43+;;;;;;;;;;/m1........../s1

InChI key

WSDSONZFNWDYGZ-BKFDUBCBSA-I

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Colistin sodium methanesulfate is used to permeabilize bacterial cell membranes and to study mannose-resistant haemagglutination.

Biochem/physiol Actions

Colistin is a cationic, multicomponent lipopeptide antibiotic. It consists of a cyclic heptapeptide with tripeptide side chain, acylated at the N terminus by a fatty acid. It exhibits a potent anti-endotoxin. Colistin interacts with the anionic LPS molecules and displaces calcium and magnesium ions. It solubilzes the cytoplasmic membrane resulting in leakage of cytosol which contributes to the bactericidal effect.
Mode of Action: Binds to lipids on the cell cytoplasmic membrane of Gram-negative bacteria and disrupts the cell wall integrity.
Antimicrobial spectrum: Gram-negative bacteria.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachin Gupta et al.
Indian journal of critical care medicine : peer-reviewed, official publication of Indian Society of Critical Care Medicine, 13(2), 49-53 (2009-11-03)
One of the greatest achievements of modern medicine is the development of antibiotics against life-threatening infections, but the emergence of multidrug-resistant (MDR) gram negative bacteria has drastically narrowed down the therapeutic options against them. This limitation has led clinicians to
Matthew E Falagas et al.
Expert review of anti-infective therapy, 6(5), 593-600 (2008-10-14)
Despite the constantly increasing need for new antimicrobial agents, antibiotic drug discovery and development seem to have greatly decelerated in recent years. Presented with the significant problem of advancing antimicrobial resistance, the global scientific community has attempted to find alternative
Roberto Imberti et al.
Chest, 138(6), 1333-1339 (2010-06-19)
Infections caused by multidrug-resistant gram-negative bacteria have caused a resurgence of interest in colistin. To date, information about pharmacokinetics of colistin is very limited in critically ill patients, and no attempts have been made to evaluate its concentration in BAL.
M.J. Rogers et al.
Infection (Munich), 14, 79-79 (1986)
S M Garonzik et al.
Antimicrobial agents and chemotherapy, 55(7), 3284-3294 (2011-05-11)
With increasing clinical emergence of multidrug-resistant Gram-negative pathogens and the paucity of new agents to combat these infections, colistin (administered as its inactive prodrug colistin methanesulfonate [CMS]) has reemerged as a treatment option, especially for critically ill patients. There has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service