Skip to Content
Merck
All Photos(3)

Key Documents

M14102

Sigma-Aldrich

Diethylene glycol dimethyl ether

ReagentPlus®, 99%

Synonym(s):

2-Methoxyethyl ether, Bis(2-methoxyethyl) ether, Dimethyldiglycol, ‘Diglyme’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2CH2)2O
CAS Number:
Molecular Weight:
134.17
Beilstein:
1736101
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.6 (vs air)

Quality Level

vapor pressure

3 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

370 °F

expl. lim.

17.4 %

color

APHA: ≤10

refractive index

n20/D 1.408 (lit.)

pH

7 (20 °C)

bp

162 °C (lit.)

mp

−64 °C (lit.)

density

0.944 g/mL at 20 °C (lit.)
0.939 g/mL at 25 °C (lit.)

SMILES string

COCCOCCOC

InChI

1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3

InChI key

SBZXBUIDTXKZTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylene glycol dimethyl ether (diglyme), a hydrophilic ether, is an electron-donor solvent. Its toxicity has been assessed. The effect of complexation of diglyme with lithium trifluoromethanesulfonate on the conformation has been investigated. It is reported to enhance the ability of KF-MeOH-Al2O3 (potassium fluoride-methanol-alumina) reagent to replace halogen with methoxy groups in 5,8-dihydroxy-2,3-dichloro-1,4-naphthoquinones.

Application

Diethylene glycol dimethyl ether may be used as a solvent to form a solution of sodium pentaphosphacyclopentadienide.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new method for the preparation of solution of sodium pentaphosphacyclopentadienide.
Milyukov VA, et al.
Russian Chemical Bulletin, 55(7), 1297-1299 (2006)
C J Price et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 8(1), 115-126 (1987-01-01)
Diethylene glycol dimethyl ether (diEGdiME) is structurally related to several compounds which produce reproductive and developmental toxicity, including teratogenicity in laboratory animals. In the present study, diEGdiME (0, 62.5, 125, 250, or 500 mg/kg/day) was administered by gavage in distilled
Catalytic activity of diethylene glycol dimethyl ethers in conversion of 6,7-dichloro-3-ethyl-2-ethoxynaphthazarine into echinochrome trimethyl ether.
Kochergina TY and Anufriev VF.
Russ. J. Org. Chem., 38(4), 534-537 (2002)
Conformational changes in diethylene glycol dimethyl ether and poly (ethylene oxide) induced by lithium ion complexation.
Frech R and Huang W.
Macromolecules, 28(4), 1246-1251 (1995)
Simultaneous determination of six hydrophilic ethers at trace levels using coconut charcoal adsorbent and gas chromatography/mass spectrometry.
Stepien DK and Puttmann W.
Analytical and Bioanalytical Chemistry, 405(5), 1743-1751 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service