Skip to Content
Merck
All Photos(4)

Key Documents

E27408

Sigma-Aldrich

1,2-Dimethoxyethane

ReagentPlus®, ≥99%, inhibitor-free

Synonym(s):

mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1209237
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
bp:
85 °C (lit.)
vapor pressure:
48 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.1 (20 °C, vs air)

Quality Level

vapor pressure

48 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

396 °F

expl. lim.

10.4 %

impurities

≤0.5% (water)

refractive index

n20/D 1.379 (lit.)

pH

7

bp

85 °C (lit.)

mp

−58 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

SMILES string

COCCOC

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dimethoxyethane (DME) is an inert aprotic polar solvent for electrolytes of lithium batteries, polysilicones, oligo- and polysaccharides.

Application

1,2-Dimethoxyethane can be used as a solvent to synthesize:
  • Oxazoles from α-acylamino ketones via Robinson-Gabriel reaction.
  • 5-Arylhistidines via Suzuki-Miyaura cross-coupling between 5-bromohistidine and arylboronic acids.
  • Aryl thioethers via Pd-catalyzed thioetherification of aryl halides with aliphatic and aromatic thiols.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Organometallics, 13, 1498-1498 (1994)
Tetrahedron Letters, 34, 4591-4591 (1993)
Inorganic Chemistry, 33, 1685-1685 (1994)
Peter A Campochiaro et al.
Ophthalmology, 122(3), 545-554 (2014-12-03)
AKB-9778 is a small-molecule competitive inhibitor of vascular endothelial-protein tyrosine phosphatase (VE-PTP) that promotes Tie2 activation and reduces vascular leakage and neovascularization in mouse models. The purpose of this study was to test the safety, tolerability, pharmacokinetics, and biological activity
Andrew Martins et al.
Organic letters, 12(22), 5186-5188 (2010-10-19)
A palladium-catalyzed crossed biaryl coupling/reduction sequence enables the formation of meta-substituted biaryls via solvent-mediated arylpalladium(II) reduction. Isotope labeling studies determined that the decomposition of 1,2-dimethoxyethane (DME) is indeed involved in the reductive process.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service