Skip to Content
Merck
All Photos(3)

Key Documents

271020

Sigma-Aldrich

1,3-Dioxolane

anhydrous, contains ~75 ppm BHT as inhibitor, 99.8%

Synonym(s):

Ethylene glycol methylene ether, Formaldehyde ethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Beilstein:
102453
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor density

2.6 (vs air)

vapor pressure

70 mmHg ( 20 °C)

Assay

99.8%

form

liquid

autoignition temp.

525 °F

contains

~75 ppm BHT as inhibitor

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.401 (lit.)

bp

75-76 °C/1.013 hPa

mp

−95 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

C1COCO1

InChI

1S/C3H6O2/c1-2-5-3-4-1/h1-3H2

InChI key

WNXJIVFYUVYPPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-dioxolane is a heterocyclic acetal that is commonly used as a green solvent in formulations and production processes. It is also used as a reactant in organic synthesis. Long-term storage of 1,3-dioxolane forms organic peroxides. This process can be suppressed by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT removes the free radicals required for peroxide formation.

Application

1,3-dioxolane can be used as a solvent:      
  • For the extraction of biodegradable poly(3-hydroxybutyrate) (PHB) from biomass.     
  • To produce stable dispersions of multiwalled carbon nanotubes (MWCNTs) for the synthesis of modified glassy carbon electrodes.
  • To synthesize copolymers (polyoxymethylene) by cationic copolymerization of 1,3,5-trioxane in the presence of maghnite-H+ as a catalyst.

It is also used as a reactant to prepare quasi-solid-state poly(1,3-dioxolane) electrolyte by Sc(OTf)3 catalyzed ring-opening polymerization reactions.
      

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binary electrolyte based on tetra (ethylene glycol) dimethyl ether and 1, 3-dioxolane for lithium-sulfur battery.
Chang DR, et al.
Journal of Power Sources, 112(2), 452-460 (2002)
1, 3-Dioxolane: A green solvent for the preparation of carbon nanotube-modified electrodes.
Moscoso R, et al.
Electrochemical Communications, 48, 69-72 (2014)
Chemical synthesis of heterocyclic-sugar nucleoside analogues.
Giovanni Romeo et al.
Chemical reviews, 110(6), 3337-3370 (2010-03-18)
Lukas J Goossen et al.
The Journal of organic chemistry, 73(21), 8631-8634 (2008-10-23)
An efficient synthesis of the angiotensin II receptor antagonist telmisartan is presented involving a decarboxylative cross-coupling of isopropyl phthalate (1) with 2-(4-chlorophenyl)-1,3-dioxolane (2c) as the key step (85% yield). The benzimidazole moiety is constructed regioselectively via a reductive amination-condensation sequence
Dong Zhou et al.
Nuclear medicine and biology, 35(6), 655-663 (2008-08-06)
Progesterone receptors (PRs) are present in many breast tumors, and their levels are increased by certain endocrine therapies. They can be used as targets for diagnostic imaging and radiotherapy. 16alpha,17alpha-[(R)-1'-alpha-(5-[(76)Br]Bromofurylmethylidene)dioxyl]-21-hydroxy-19-norpregn-4-ene-3,20-dione ([(76)Br]16alpha,17alpha-[(R)-1'-alpha-(5-bromofurylmethylidene)dioxyl]-21-hydroxy-19-norpregn-4-ene-3,20-dione (3)), a PR ligand with relative binding affinity (RBA)=65

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service