Skip to Content
Merck
All Photos(2)

Key Documents

8.00856

Sigma-Aldrich

Ethylene glycol dimethyl ether

(stabilised) for synthesis

Synonym(s):

1,2-Dimethoxyethane, mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1209237
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-794-9
NACRES:
NA.22
Assay:
≥99.9% (GC)
bp:
85 °C (lit.)
vapor pressure:
48 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.1 (20 °C, vs air)

Quality Level

vapor pressure

48 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

396 °F

potency

2000 mg/kg LD50, skin (Rabbit)

expl. lim.

10.4 %

refractive index

n20/D 1.379 (lit.)

pH

7

kinematic viscosity

0.455 cSt(25 °C)

bp

85 °C (lit.)

mp

−58 °C (lit.)

solubility

soluble 1000 g/L

density

0.867 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

COCCOC

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol dimethyl ether, also known as dimethoxyethane, has gained significant attention in recent years due to its favorable characteristics, including a high energy density and cetane number. It possesses excellent solubility properties and is used as an environmentally friendly solvent and effective etherification agent in industries such as cosmetics, perfumes, and pharmaceuticals. Notably, it finds extensive application in batteries and electrolytes due to its desirable properties.[1]

Application

Ethylene glycol dimethyl ether can be used as a:
  • Co-solvent in the synthesis of Tris(trimethylsilyl)phosphine (P(SiMe3)3).[2]
  • Solvent in the tailored mono carborane electrolyte solution of Ca(CB11H12)2, which was utilized in the development of Ca metal batteries.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective synthesis of dimethoxyethane via directly catalytic etherification of crude ethylene glycol
Yu W, et al.
Green Chemistry, 19(14), 3327-3333 (2017)
Role of acid in precursor conversion during InP quantum dot synthesis
Gary DC, et al.
Chemistry of Materials, 25(12), 2463- 2469 (2013)
Calcium Metal Batteries with Long Cycle Life Using a Hydride-Based Electrolyte and Copper Sulfide Electrode
Advanced Science Letters, 10(22), 2301178-2301178 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service