C5510
Activated charcoal
acid-washed with phosphoric and sulfuric acids
Synonym(s):
Charcoal activated
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52
Recommended Products
vapor pressure
<0.1 mmHg ( 20 °C)
Quality Level
form
powder
autoignition temp.
842 °F
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
resistivity
1375 μΩ-cm, 20°C (graphite)
impurities
≤1.0% acid solubles
≤10% moisture content
mp
3550 °C (lit.)
greener alternative category
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.
Application
Activated charcoal was used to adsorb lignin and hemicelluloses in spent liquor from neutral sulfite semichemical (NSSC) pulping process.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mehdi Dashtban et al.
Journal of environmental management, 136, 62-67 (2014-02-26)
Hemicelluloses and lignin present in the spent liquor (SL) of neutral sulfite semichemical (NSSC) pulping process can potentially be converted into value-added products such as furfural, hydroxymethylfurfural, levulinic acid, phenols and adhesives. However, the direct conversion of hemicelluloses and lignin
Yin Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 533-538 (2013-12-25)
Stomatal pores surrounded by a pair of guard cells in the plant epidermis control gas exchange between plants and the atmosphere in response to light, CO2, and the plant hormone abscisic acid. Light-induced stomatal opening is mediated by at least
Kohsuke Ohmatsu et al.
Nature chemistry, 6(1), 47-51 (2013-12-19)
The development of a general catalytic method for the direct and stereoselective construction of contiguous all-carbon quaternary stereocentres remains a formidable challenge in chemical synthesis. Here, we report a highly enantio- and diastereoselective [3+2] annulation reaction of 5-vinyloxazolidinones and activated
Nathalie Butt et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 936-943 (2013-07-20)
A typical way to quantify aboveground carbon in forests is to measure tree diameters and use species-specific allometric equations to estimate biomass and carbon stocks. Using "citizen scientists" to collect data that are usually time-consuming and labor-intensive can play a
Isabelle Aillaud et al.
Dalton transactions (Cambridge, England : 2003), (44)(44), 5105-5118 (2007-11-07)
The catalysed enantioselective formation of carbon-nitrogen bonds by the hydroamination reaction is reviewed. All examples deal with substrates containing non-activated carbon-carbon multiple bonds which are transformed either via intramolecular or intermolecular reactions. Structurally different complexes already provided nitrogen containing compounds/heterocycles
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service