Skip to Content
Merck
All Photos(1)

Documents

C75004

Sigma-Aldrich

5-Cholesten-3-one

Synonym(s):

3-Keto-5-cholestene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

125-127 °C (lit.)

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,22-25H,6-8,10-17H2,1-5H3/t19-,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

GGCLNOIGPMGLDB-GYKMGIIDSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I J Kass et al.
Biochemistry, 37(51), 17990-18000 (1999-01-28)
Cholesterol oxidase catalyzes the oxidation and isomerization of cholesterol to cholest-4-en-3-one via cholest-5-en-3-one. It has been proposed that His447 acts as the general base catalyst for oxidation, and that the resulting imidazolium ion formed acts as an electrophile for isomerization.
I J Kass et al.
Bioorganic & medicinal chemistry letters, 8(19), 2663-2668 (1999-01-05)
Cholesterol oxidase stereospecifically isomerizes cholest-5-en-3-one to cholest-4-en-3-one. When the base catalyst for isomerization, Glu361, is mutated to Asp, the rate of deprotonation of cholest-5-en-3-one is not affected, but protonation of the dienolic intermediate becomes rate-limiting. This may be a consequence
J P Slotte et al.
Biochimica et biophysica acta, 1145(2), 243-249 (1993-02-09)
In this study we have examined the cholesterol oxidase (Streptomyces cinnamomeus) catalyzed conversion of either 5-cholesten-3 beta-ol or 5-cholesten-3-one into 4-cholesten-3-one in pure sterol or mixed phospholipid-containing monolayers at the air/buffer interface. The mean molecular area requirement of 5-cholesten-3-one in
Hui-Min Qin et al.
Protein expression and purification, 139, 1-7 (2017-07-18)
Cholesterol oxidases, which catalyze the degradation of cholesterol to cholest-4-en-3-one, are widely used in the pharmaceutical and food processing industries. The cholesterol oxidase from Pimelobacter simplex (PsChO3) was transformed into E. coli BL21(DE3), but it was expressed mainly as inclusion bodies
Khondker Rufaka Hossain et al.
Biochimica et biophysica acta. General subjects, 1863(8), 1243-1253 (2019-05-11)
Sterols have been reported to modulate conformation and hence the function of several membrane proteins. One such group is the Chloride Intracellular Ion Channel (CLIC) family of proteins. The CLIC protein family consists of six evolutionarily conserved protein members in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service