Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Activated charcoal
Synonym(s):
Charcoal activated, Activated charcoal
Empirical Formula (Hill Notation):
C
CAS Number:
7440-44-0
Molecular Weight:
12.01
EC Number:
231-153-3
Products
Brand
Millipore (1)
Sigma-Aldrich (10)
Supelco (1)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Technique
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
242276
Activated charcoal,
DARCO
®
, −100 mesh particle size, powder
Expand
View Pricing
161551
Activated charcoal,
powder, -100 particle size (mesh), decolorizing
Expand
View Pricing
242268
Activated charcoal,
DARCO
®
, 20-40 mesh particle size, granular
Expand
View Pricing
C2764
Activated charcoal,
untreated, granular, ≤5 mm
Expand
View Pricing
242233
Activated charcoal,
DARCO
®
, 4-12 mesh particle size, granular
Expand
View Pricing
C4386
Activated charcoal,
acid-washed with hydrochloric acid
Expand
View Pricing
05105
Activated charcoal,
powder
Expand
View Pricing
242241
Activated charcoal,
DARCO
®
, 12-20 mesh, granular
Expand
View Pricing
C7606
Activated charcoal,
meets USP testing specifications
Expand
View Pricing
C2889
Activated charcoal,
untreated, granular, 8-20 mesh
Expand
View Pricing
329428
Activated charcoal,
Norit
®
ROW 0.8 SUPRA, pellets
Expand
View Pricing
C5510
Activated charcoal,
acid-washed with phosphoric and sulfuric acids
Expand
View Pricing
C3014
Activated charcoal,
untreated, granular, 20-60 mesh
Expand
View Pricing
C9157
Activated charcoal,
suitable for cell culture, suitable for plant cell culture
Expand
View Pricing
05112
Activated charcoal,
suitable for GC
Expand
View Pricing