Skip to Content
Merck
All Photos(9)

Key Documents

746436

Sigma-Aldrich

Potassium chloride

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.005% Insoluble matter

pH

5.4-8.6 (25 °C, 5% in solution)

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤0.01%
chlorate, nitrate (as NO3-): ≤0.003%
iodide (I-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.001%

cation traces

Ba:, passes test (limit about 0.001%)
Ca: ≤0.002%
Fe: ≤3 ppm
Mg: ≤0.001%
Na: ≤0.005%
heavy metals (as Pb): ≤5 ppm

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium chloride is an inorganic salt commonly used in fertilizer industry. Its generation via chlorination of microcline has been reported. The intrinsic and extrinsic conductivity of KCl crystals have been measured. The impact of KCl on the activity of mitochondrial membrane, cristae and matrix proteins has been investigated. It facilitates the preparation of isoxazolines in aqueous medium.

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.
Potassium chloride may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

It has been used in the preparation of the following:
  • Amorphous calcium phosphate (ACP) deposition solution, used for ACP coating on Ti coupons
  • Modified methylene blue (MB) solution, utilized in the MB two-phase titration method
  • 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid (HEPES)-containing artificial cerebrospinal fluid (ACSF) solutions
  • Artificial sea water

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lan Lin et al.
PLoS pathogens, 15(6), e1007795-e1007795 (2019-06-07)
Infection with the Streptococcus suis (S. suis) epidemic strain can cause Streptococcal toxic shock-like syndrome (STSLS), which is characterized by a cytokine storm, dysfunction of multiple organs and a high incidence of mortality despite adequate treatment. Despite some progress concerning
Hanne Sørup Tastesen et al.
Amino acids, 46(7), 1659-1671 (2014-03-25)
High-protein diets induce alterations in metabolism that may prevent diet-induced obesity. However, little is known as to whether different protein sources consumed at normal levels may affect diet-induced obesity and associated co-morbidities. We fed obesity-prone male C57BL/6J mice high-fat, high-sucrose
Sandra S Wise et al.
Toxicology and applied pharmacology, 279(2), 113-118 (2014-06-22)
Sea turtles are a charismatic and ancient ocean species and can serve as key indicators for ocean ecosystems, including coral reefs and sea grass beds as well as coastal beaches. Genotoxicity studies in the species are absent, limiting our understanding
Wayne O Miles et al.
Molecular and cellular biology, 35(24), 4199-4211 (2015-10-07)
The lysine (K)-specific demethylase (LSD1) family of histone demethylases regulates chromatin structure and the transcriptional potential of genes. LSD1 is frequently deregulated in tumors, and depletion of LSD1 family members causes developmental defects. Here, we report that reductions in the
Qing-Fu Cao et al.
Molecular and cellular biology, 35(20), 3459-3470 (2015-07-29)
Restores TBP function 1 (Rtf1) is generally considered to be a subunit of the Paf1 complex (PAF1C), a multifunctional protein complex involved in histone modification and transcriptional or posttranscriptional regulation. Rtf1, however, is not stably associated with the PAF1C in

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service