Skip to Content
Merck
All Photos(8)

Key Documents

792519

Sigma-Aldrich

Sodium bicarbonate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%

Synonym(s):

Sodium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for EPA 1621
suitable for EPA 300

product line

Redi-Dri

Assay

≥99.7%

form

powder

quality

free-flowing

impurities

≤0.015% insoluble matter

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

density

2.16 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
phosphate (PO43-): ≤0.001 ppm
sulfur compounds (as SO42-): ≤0.003%

cation traces

Ca: ≤0.02%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.005%
NH4+: ≤5 ppm
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bicarbonate is commonly known as baking soda. It serves as a source of carbon dioxide and is used to prepare a variety of sodium salts. It is also an important pH-regulating agent.

Application

Sodium bicarbonate can be used as a reactant to synthesize monosodium Schiff bases by the condensation reaction of aldehydes and amino acids.

Features and Benefits

  • Increased efficiency – Eliminates time and effort of loosening hardened powders      
  • Safety – Eliminates tools of force used to break up clumps
  • Ecology – Reduces waste as it does not stick to the packaging
  • Economy – Faster preparation and solvation increase productivity and reduce costs     
  • Assured Quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Liang et al.
Electrophoresis, 36(5), 731-736 (2014-11-26)
Although the existing theories have predicted enhancement of electrophoretic mobility of microparticles near a solid wall, the relevant experimental studies are rare. This is mainly due to difficulties in experimentally controlling and measuring particle-wall separations under dynamic electrophoretic conditions. This
Joshua C Tremblay et al.
The Journal of physiology, 593(3), 723-737 (2014-11-25)
Blood flow through intrapulmonary arteriovenous anastomoses (IPAVA) is increased by acute hypoxia during rest by unknown mechanisms. Oral administration of acetazolamide blunts the pulmonary vascular pressure response to acute hypoxia, thus permitting the observation of IPAVA blood flow with minimal
Kate M Hatzopoulos et al.
Investigative ophthalmology & visual science, 55(6), 3957-3970 (2014-06-05)
Retinal angiogenesis, neural dysfunction, and microglial activation occur in both humans with retinopathy of prematurity and in animal models of the disease. The aim of this study was to assess whether blockade of the renin-angiotensin system ameliorated these effects in
Roberto J Diaz et al.
American journal of physiology. Cell physiology, 306(12), C1191-C1199 (2014-04-25)
We have previously shown that ischemic preconditioning (IPC) protection against necrosis in whole hearts and in both fresh and cultured cardiomyocytes, as well as the improved regulatory volume decrease to hypoosmotic swelling in cardiomyocytes, is abrogated through Cl(-) channel blockade
Erik van Kampen et al.
Journal of leukocyte biology, 96(5), 833-841 (2014-07-16)
Alterations in DNA methylation patterns in peripheral blood leukocytes precede atherosclerotic lesion development in mouse models of atherosclerosis and have been linked to cardiovascular death in patients. The aim of this study is to investigate the long-term changes induced by

Articles

JosiPhos CyPF-tBu and palladium give catalyst for alkoxylation of activated heteroaryl halides with primary, secondary, and tertiary alcohols

Effective in key synthesis reactions like Knoevenagel condensation, thalidomide synthesis, and Suzuki coupling for sustainable chemical transformations.

Effective in key synthesis reactions like Knoevenagel condensation, thalidomide synthesis, and Suzuki coupling for sustainable chemical transformations.

Effective in key synthesis reactions like Knoevenagel condensation, thalidomide synthesis, and Suzuki coupling for sustainable chemical transformations.

See All

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service