Skip to Content
Merck
All Photos(4)

Key Documents

P3911

Sigma-Aldrich

Potassium chloride

ACS reagent, 99.0-100.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder

grade

ACS reagent

Quality Level

Assay

99.0-100.5%

form

powder

impurities

≤0.005% Insoluble matter

pH

5.4-8.6 (25 °C, 5% in solution)

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤0.01%
chlorate, nitrate (as NO3-): ≤0.003%
iodide (I-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.001%

cation traces

Ba:, passes test (limit about 0.001%)
Ca: ≤0.002%
Fe: ≤3 ppm
Mg: ≤0.001%
Na: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES )

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium chloride is an inorganic salt commonly used in fertilizer industry. Its generation via chlorination of microcline has been reported. The intrinsic and extrinsic conductivity of KCl crystals have been measured. The impact of KCl on the activity of mitochondrial membrane, cristae and matrix proteins has been investigated. It facilitates the preparation of isoxazolines in aqueous medium.

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.
Potassium chloride has been used in the preparation of the following:
  • Amorphous calcium phosphate (ACP) deposition solution, used for ACP coating on Ti coupons.
  • Modified methylene blue (MB) solution, utilized in the MB two-phase titration method.
  • 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid (HEPES)-containing artificial cerebrospinal fluid (ACSF) solutions.
  • Artificial sea water.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Comparison of titanium soaked in 5M NaOH or 5M KOH solutions.
Kim C, et al.
Materials Science and Engineering, C, 33(1), 327-339 (2013)
Stability of stearic acid monolayers on Artificial Sea Water.
Brzozowska AM, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 407, 38-48 (2012)
An environmentally benign synthesis of isoxazolines and isoxazoles mediated by potassium chloride in water.
Han L, et al.
Tetrahedron Letters, 55(14), 2308-2311 (2014)
Leyu Cui et al.
Analytical chemistry, 86(22), 11055-11061 (2014-11-05)
The methylene blue (MB) two-phase titration method is a rapid and efficient method for determining the concentrations of anionic surfactants. The point at which the aqueous and chloroform phases appear equally blue is called Epton's end point. However, many inorganic
Suman Mishra et al.
International journal of proteomics, 2015, 647408-647408 (2016-01-20)
Potassium chloride (KCl) has been commonly used in homogenization buffer and procedures of protein extraction. It is known to facilitate release of membrane-associated molecules but the higher concentration of KCl may affect the integrity of mitochondria by breaching the electrostatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service