Skip to Content
Merck
All Photos(3)

Key Documents

307432

Sigma-Aldrich

1,2-Dimethoxyethane

suitable for HPLC, 99.9%, inhibitor-free

Synonym(s):

mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1209237
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.1 (20 °C, vs air)

vapor pressure

48 mmHg ( 20 °C)

Assay

99.9%

form

liquid

autoignition temp.

396 °F

purified by

glass distillation

expl. lim.

10.4 %

technique(s)

HPLC: suitable

impurities

<0.030% water

refractive index

n20/D 1.379 (lit.)

pH

~7

bp

85 °C (lit.)

mp

−58 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 220 nm Amax: 1.00
λ: 250 nm Amax: 0.20
λ: 300 nm Amax: 0.03
λ: 350-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

COCCOC

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This solvent is glass distilled, submicron filtered and subjected to rigorous specification testing to provide lot-to-lot consistency.

Application

Designed for use with HPLC instrumentation and organic synthesis applications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

NMR Chemical Shifts of Common Laboratory Solvents as Trace Impurities.
Hugo E. Gottlieb et al.
The Journal of organic chemistry, 62(21), 7512-7515 (2001-10-24)
Theoretical study of cation/ether complexes: Alkali metal cations with 1, 2-dimethoxyethane and 12-crown-4.
Hill SE, et al.
The Journal of Physical Chemistry A, 102(21), 3813-3819 (1998)
The catalytic Pauson-Khand reaction promoted by a small amount of 1, 2-dimethoxyethane or water.
Sugihara T and Yamaguchi M.
Synlett, 12, 1384-1386 (1998)
Taeyun Ku et al.
Nature methods, 17(6), 609-613 (2020-05-20)
We developed entangled link-augmented stretchable tissue-hydrogel (ELAST), a technology that transforms tissues into elastic hydrogels to enhance macromolecular accessibility and mechanical stability simultaneously. ELASTicized tissues are highly stretchable and compressible, which enables reversible shape transformation and faster delivery of probes
Eiron Cudaback et al.
Glia, 63(1), 51-65 (2014-08-06)
Apolipoprotein E (apoE) is well known as a regulator of cholesterol homeostasis, and is increasingly recognized to play a prominent role in the modulation of innate immune response, including cell-to-cell communication and migration. Alzheimer's disease (AD) is a slowly progressive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service