Skip to Content
Merck
All Photos(1)

Key Documents

261734

Sigma-Aldrich

Petroleum ether

spectrophotometric grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.07

grade

spectrophotometric grade

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.020% water

evapn. residue

<0.0005%

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)

density

0.64 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 210 nm Amax: 1.00
λ: 220 nm Amax: 0.30
λ: 230 nm Amax: 0.10
λ: 240 nm Amax: 0.05
λ: 260-400 nm Amax: 0.01

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid chromatographic technique for preparative separations with moderate resolution.
Still WC, et al.
The Journal of Organic Chemistry, 43(14), 2923-2925 (1978)
S E Besra et al.
Journal of ethnopharmacology, 54(1), 1-6 (1996-10-01)
Pharmacological studies were conducted with the petroleum ether extract of leaves of the plant Litchi chinensis Gaertn. (Sapindaceae) on experimental animals. The extract was found to possess antiinflammatory, analgesic and antipyretic activity. Acute toxicity studies revealed that the extract, up
S Edelfors et al.
Pharmacology & toxicology, 84(5), 197-200 (1999-06-11)
Female rats were exposed to white spirit (400 and 800 ppm for 6 hr/day) at day 7-20 during pregnancy. Thirty-five days after birth all female offspring were sacrificed, the brains removed, and the synaptosomal fractions prepared for in vitro studies.
Gunnar Damgård Nielsen et al.
Basic & clinical pharmacology & toxicology, 98(2), 115-123 (2006-02-01)
Numerous studies have suggested that long-term occupational exposure to white spirit may cause chronic toxic encephalopathy (WHO 1996). This review summarizes the chronic nervous system effects of white spirit in animal studies during a 30-year period. First, routine histopathology was
H R Lam et al.
Toxicology letters, 80(1-3), 39-48 (1995-10-01)
The effects of 3 weeks' or 6 months' inhalation exposure of rats to aromatic white spirit 6 h/day, 5 days/week at 0, 400, or 800 ppm were studied. Synaptosomal neurochemistry was investigated as index of the in situ conditions in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service