Skip to Content
Merck
All Photos(4)

Key Documents

1.01769

Supelco

Petroleum ether

for denaturation

Synonym(s):

Light Hydrocarbon Solvent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12191502
NACRES:
NB.21
bp:
>50 °C/1013 hPa
vapor pressure:
400 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

400 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

240 °C

expl. lim.

1-7.4 % (v/v)

bp

>50 °C/1013 hPa

transition temp

flash point -31 °C

density

0.666 g/cm3 at 15 °C

storage temp.

2-30°C

Application


  • Investigation on Compositional Structures of the Organic Matter and Biomarker Distributions in Shengli Lignite.: This study delves into the organic matter′s compositional structures and biomarker distributions in Shengli lignite, revealing insights relevant to petrochemical applications, including the use of solvents like petroleum ether (Liu et al., 2024).

  • Di-tert-butylphosphate Derived Thermolabile Calcium Organophosphates: Precursors for Ca(H2PO4)2, Ca(HPO4), α-/β-Ca(PO3)2, and Nanocrystalline Ca10(PO4)6(OH)2.: Research on calcium organophosphates, detailing the role of petroleum ether in preparing samples for structural analysis, highlighting its utility in materials science and engineering (Verma and Murugavel, 2020).

Analysis Note

Density (d 20 °C/20 °C): 0.600 - 0.700
Miscibility with ethanol: conforms
Miscibility with water: conforms
Course of the distillation analysis: conforms
Petroleum ether has been tested as denaturant by authorities.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-23.8 °F

Flash Point(C)

-31 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service