Skip to Content
Merck
All Photos(2)

Key Documents

184519

Sigma-Aldrich

Petroleum ether

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.07
grade:
ACS reagent
bp:
30-60 °C (lit.)
35-60 °C
vapor pressure:
25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

grade

ACS reagent

Quality Level

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

dilution

(for analytical testing)

impurities

acidity, passes test

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)
35-60 °C

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Petroleum ether is a colorless liquid. Extracts of leaves of Litchi chinensis Gaertn. (Sapindaceae) have been prepared in petroleum ether and were evaluated for antiinflammatory, analgesic and antipyretic activities. Pentanes and hexanes are the major constituents of petroleum ether.
Petroleum ether is a mixture of low boiling aliphatic hydrocarbons (mainly hexane and pentane) that is generally used as a laboratory solvent.

Application

Petroleum ether has been used for the analysis of total dietary fiber content by employing a total dietary fibre assay kit. It may be used in the absorption chromatographic purification of organic compounds.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F

Flash Point(C)

-49 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Lide DR
CRC Handbook of Chemistry and Physics, 85 (2004)
Eagleson M.
Concise Encyclopedia Chemistry, 783-783 (1994)
S E Besra et al.
Journal of ethnopharmacology, 54(1), 1-6 (1996-10-01)
Pharmacological studies were conducted with the petroleum ether extract of leaves of the plant Litchi chinensis Gaertn. (Sapindaceae) on experimental animals. The extract was found to possess antiinflammatory, analgesic and antipyretic activity. Acute toxicity studies revealed that the extract, up
P S Tofts et al.
Magnetic resonance imaging, 15(1), 69-75 (1997-01-01)
The selection of a suitable low dielectric constant liquid for radiofrequency coil nonuniformity measurements is described. Measurements of dielectric constant (relative permittivity) were made on a range of candidate liquids. After excluding liquids that were too inflammable or too viscous
S Dauger et al.
Archives de pediatrie : organe officiel de la Societe francaise de pediatrie, 10(3), 221-223 (2003-06-28)
The Authors report a case of acute White-Spirit poisoning with pulmonary hypertension associated to respiratory distress syndrome. A 14-month-old infant drank an unknown quantity of White-Spirit while his parents were painting. After he spontaneously vomited, he presented a seizure at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service