Skip to Content
Merck
All Photos(4)

Key Documents

228710

Sigma-Aldrich

Sodium nitroferricyanide(III) dihydrate

ACS reagent, ≥99%

Synonym(s):

Nitroprusside sodium, SNP, Sodium nitroprusside, Sodium pentacyanonitrosylferrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
297.95
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
grade:
ACS reagent
form:
crystals
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥99%

form

crystals

reaction suitability

reagent type: catalyst
core: iron

impurities

≤0.01% insolubles

anion traces

chloride (Cl-): ≤0.02%
sulfate (SO42-): passes test (lim. ~1%)

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O=N[Fe--](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q;;;;;2*-1;2*+1;;

InChI key

OIRZWVYIQXBRFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium nitroferricyanide(III) dihydrate (sodium nitroprusside dihydrate, SNP) is a dihydrate form of sodium nitroprusside. It is comprised of ferrous iron complexed with nitric oxide (NO) and five cyanide ions. It serves as a donor of nitric oxide.

Application

Sodium nitroferricyanide(III) dihydrate can be used as a reagent to produce nitric oxide radicals (NO) via UV radiation-induced photolysis. These radicals are employed for the construction of amino acid-based supramolecular hydrogels, which find potential applications in disease diagnosis and therapy including bioimaging, bio-detection, therapeutic delivery, and tissue engineering. Additionally, SNP also serves as a catalyst in the Phenate Method for the quantitative measurement of ammonia in water samples.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyhedron, 22, 2527-2535 (2003)
Anal. Profiles Drug Subst., 6, 487-513 (1977)
Oluwatobiloba Osikoya et al.
The Journal of physiology, 597(15), 3833-3852 (2019-06-06)
In vivo, uterine perivascular adipose tissue (PVAT) potentiates uterine artery blood flow in pregnant rats, although not in non-pregnant rats. In isolated preparations, uterine PVAT has pro-contractile and anti-dilatory effects on uterine arteries. Pregnancy induces changes in uterine arteries that
Milos R Filipovic et al.
Journal of medicinal chemistry, 56(4), 1499-1508 (2013-02-20)
Hydrogen sulfide (H2S) has been increasingly recognized as an important signaling molecule that regulates both blood pressure and neuronal activity. Attention has been drawn to its interactions with another gasotransmitter, nitric oxide (NO). Here, we provide evidence that the physiological
Song Guo et al.
Cephalalgia : an international journal of headache, 33(5), 301-307 (2013-02-14)
Sodium nitroprusside (SNP) is a powerful vasodilatory agent that, similarly to glyceryl trinitrate (GTN), releases nitric oxide (NO) but in contrast does not pass the blood-brain barrier. Nevertheless, it has already been used in animal models without any knowledge of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service