Skip to Content
Merck
All Photos(2)

Key Documents

1.05614

Supelco

Sodium hypochlorite solution

(6-14% active chlorine) EMPLURA®

Synonym(s):

Sodium hypochlorite solution, Soda bleaching lye, Eau de Labarraque

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO
CAS Number:
Molecular Weight:
74.44
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
form:
liquid

Agency

suitable for SM 4500 - NH3

Quality Level

vapor pressure

20 hPa ( 20 °C)

product line

EMPLURA®

form

liquid

concentration

6-14% (NaClO)

impurities

≤1.0% Free alkali (acidimetric, as NaOH)

pH

12-13 (20 °C in H2O)

bp

102 °C/1013 hPa

mp

-20 °C

density

1.22-1.25 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[Na+].[O-]Cl

InChI

1S/ClO.Na/c1-2;/q-1;+1

InChI key

SUKJFIGYRHOWBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

(6-14% active chlorine) EMPLURA®

Application


  • A Mechanistic Spectrum of O-H Bond Cleavage Observed for Reactions of Phenols with a Manganese Superoxo Complex.: This article provides a mechanistic understanding of the reactivity of phenols, such as 4-nitrophenol, with manganese superoxo complexes, potentially leading to new catalytic methods for environmental and synthetic applications (Tian et al., 2024).


Analysis Note

Assay (active chlorine; iodometric, Cl₂)6 - 14 %Free alkali (acidimetric, as NaOH)≤ 1.0 %The content stated refers to the filling day. Depending on storage period and temperature a decrease in content is to be expected

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service