Skip to Content
Merck
All Photos(1)

Documents

Y0000402

Pyrantel embonate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pyrantel pamoate, 1-Methyl-2-(2-[2-thienyl]ethenyl)-1,4,5,6-tetrahydropyrimidine compound with 4,4′-methylenebis(3-hydroxy-2-naphthoic acid), Pyrantel embonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H14N2S · C23H16O6
CAS Number:
Molecular Weight:
594.68
Beilstein:
9034755
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pyrantel

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1CCCN=C1\C=C\c2cccs2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(O)=O)c3O

InChI

1S/C23H16O6.C11H14N2S/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-13-8-3-7-12-11(13)6-5-10-4-2-9-14-10/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-6,9H,3,7-8H2,1H3/b;6-5+

InChI key

AQXXZDYPVDOQEE-MXDQRGINSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pyrantel embonate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Schwartz et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(48), E5169-E5177 (2014-11-19)
Basophils orchestrate protection against reinfections with gastrointestinal helminths and ticks, but the underlying mechanisms remain elusive. We investigated the role of Fc receptors on basophils, the antibody isotypes IgG1 and IgE, and basophil-derived IL-4/IL-13 during challenge infections with Heligmosomoides polygyrus
[The action of pyrantel pamoate in the therapy of helminth infestations].
C Fossati
La Clinica terapeutica, 93(6), 713-717 (1980-06-30)
Tara E Paterson et al.
Veterinary parasitology, 205(3-4), 687-696 (2014-09-30)
Advocate(®) (2.5% moxidectin+10% imidacloprid) (Bayer HealthCare, Leverkusen, Germany) is a multiparasiticidal spot-on authorized for treating canine demodicosis in many countries. This blinded, randomized three-phase clinical trial compared its efficacy employing different dosing regimens with that of ivermectin. In the blinded
Pyrantel pamoate for pinworm infestation.
G Van Riper
American pharmacy, NS33(2), 43-45 (1993-02-01)
Oxantel pamoate-albendazole for Trichuris trichiura infection.
Kosuke Yasukawa
The New England journal of medicine, 370(20), 1952-1952 (2014-05-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service