Skip to Content
Merck
All Photos(1)

Key Documents

A0325100

Albendazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methyl 5-(propylthio)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O2S
CAS Number:
Molecular Weight:
265.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

albendazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCSc1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

HXHWSAZORRCQMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Albendazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Adrenal gland,Blood,male reproductive organs,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peiling Yap et al.
The American journal of tropical medicine and hygiene, 89(1), 23-31 (2013-05-22)
Post-treatment soil-transmitted helminth re-infection patterns were studied as part of a randomized controlled trial among school-aged children from an ethnic minority group in Yunnan province, People's Republic of China. Children with a soil-transmitted helminth infection (N = 194) were randomly
Ruth Ann Baird et al.
Neurology, 80(15), 1424-1429 (2013-04-10)
To review the evidence base for different treatment strategies in intraparenchymal neurocysticercosis in adults and children. A literature search of Medline, EMBASE, LILACS, and the Cochrane Database from 1980 to 2008, updated in 2012, resulted in the identification of 10
Aprilianto E Wiria et al.
PloS one, 8(3), e57899-e57899 (2013-03-26)
Helminth infections are proposed to have immunomodulatory activities affecting health outcomes either detrimentally or beneficially. We evaluated the effects of albendazole treatment, every three months for 21 months, on STH, malarial parasitemia and allergy. A household-based cluster-randomized, double-blind, placebo-controlled trial
Marlieke E A de Kraker et al.
Tropical medicine & international health : TM & IH, 11(5), 718-728 (2006-04-28)
To determine the efficacies of combinations of ivermectin or diethylcarbamazine and albendazole, which are recommended for use in mass treatment programmes against lymphatic filariasis. Review of published trends in microfilarial (mf) intensities after treatment with these combination therapies. By fitting
M E Oxberry et al.
Journal of veterinary pharmacology and therapeutics, 23(3), 113-120 (2000-12-08)
Trophozoites of the protozoan parasite Giardia duodenalis were exposed to various albendazole concentrations for 4 h, washed, fixed and incubated with antibodies raised against albendazole and its two major metabolites albendazole sulphoxide and albendazole sulphone. Tubulin antibodies were also used.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service