Skip to Content
Merck
All Photos(1)

Key Documents

P2670000

Praziquantel

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(Cyclohexylcarbonyl)-1,2,3,6,7-11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2O2
CAS Number:
Molecular Weight:
312.41
Beilstein:
761557
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

praziquantel

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1CN(CC2N1CCc3ccccc23)C(=O)C4CCCCC4

InChI

1S/C19H24N2O2/c22-18-13-20(19(23)15-7-2-1-3-8-15)12-17-16-9-5-4-6-14(16)10-11-21(17)18/h4-6,9,15,17H,1-3,7-8,10-13H2

InChI key

FSVJFNAIGNNGKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Praziquantel EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Doenhoff et al.
Parasitology, 136(13), 1825-1835 (2009-03-14)
Treatment with praziquantel (PZQ) has become virtually the sole basis of schistosomiasis control in sub-Saharan Africa and elsewhere, and the drug is reviewed here in the context of the increasing rate that it is being used for this purpose. Attention
Wei Wu et al.
Parasitology research, 112(3), 909-915 (2013-01-30)
Schistosomiasis remains a major public health problem with an estimated 200 million people infected in the world, and in China, schistosomiasis japonica is endemic in the south part of the country. In 1960s, before praziquantel was developed, there were about
Jennifer Keiser et al.
PLoS neglected tropical diseases, 8(7), e2975-e2975 (2014-07-18)
Treatment and morbidity control of schistosomiasis relies on a single drug, praziquantel. Hence, there is a pressing need to develop additional therapeutics against schistosomiasis. The antimalarial drug mefloquine shows antischistosomal activity in animal models and clinical trials, which calls for
Wei Wang et al.
Parasitology research, 111(5), 1871-1877 (2012-10-12)
Since praziquantel was developed in 1970s, it has replaced other antischistosomal drugs to become the only drug of choice for treatment of human schistosomiases, due to high efficacy, excellent tolerability, few and transient side effects, simple administration, and competitive cost.
J M Bygott et al.
Acta tropica, 111(2), 95-101 (2009-04-21)
The role of praziquantel in hydatid disease has not been well defined. This review evaluates the evidence on the use of praziquantel in treatment of cystic hydatid disease from in vitro and in vivo animal studies, human clinical studies and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service