Skip to Content
Merck
All Photos(1)

Documents

46648

Supelco

Praziquantel

VETRANAL®, analytical standard

Synonym(s):

2-(Cyclohexylcarbonyl)-1,2,3,6,7-11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2O2
CAS Number:
Molecular Weight:
312.41
Beilstein:
761557
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

parasites

application(s)

clinical testing

format

neat

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

O=C1CN(CC2N1CCc3ccccc23)C(=O)C4CCCCC4

InChI

1S/C19H24N2O2/c22-18-13-20(19(23)15-7-2-1-3-8-15)12-17-16-9-5-4-6-14(16)10-11-21(17)18/h4-6,9,15,17H,1-3,7-8,10-13H2

InChI key

FSVJFNAIGNNGKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone
Praziquantel (PZQ) is a pyrazino-isoquinoline compound used as a broad-spectrum anthelmintic drug against trematode and cestode infections.

Application

PZQ may be used as a reference standard for the determination for PZQ in:
  • Surface water samples by solid phase extraction (SPE) and ultra-high performance liquid chromatography-quadrupole linear ion trap tandem mass spectrometry (UHPLC-QqLIT-MS) equipped with electrospray ionization (ESI) source.
  • Plants by HPLC with triple quadrupole mass spectrometry (HPLC-ESI-QqQ-MS/MS) operating in multiple reaction monitoring (MRM) mode.
  • Tablet formulations by reversed phase (RP) HPLC with ultraviolet (UV) detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of praziquantel phytoremediation and transformation and its removal in constructed wetland.
Marsik P, et al.
Journal of Hazardous Materials, 323, 394-399 (2017)
Analysis of anthelmintics in surface water by ultra high performance liquid chromatography coupled to quadrupole linear ion trap tandem mass spectrometry.
Zrncic M, et al.
Chemosphere, 99, 224-232 (2014)
Shona Wilson et al.
PLoS neglected tropical diseases, 7(3), e2149-e2149 (2013-04-05)
IgE specific to worm antigen (SWA) and pre-treatment eosinophil number, are associated with human immunity to re-infection with schistosomes after chemotherapeutic treatment. Treatment significantly elevates circulating IL-5 24-hr post-treatment of Schistosoma mansoni. Here we investigate if praziquantel treatment of human
Jennifer A Downs et al.
Tropical medicine & international health : TM & IH, 18(9), 1090-1096 (2013-08-14)
We explored response to single-dose praziquantel therapy in a cohort of 33 women with Schistosoma haematobium infection in rural Mwanza, Tanzania. Women with S. haematobium infection confirmed both by eggs in urine and by polymerase chain reaction (PCR) received single-dose praziquantel
A rapid stability indicating LC-method for determination of praziquantel in presence of its pharmacopoeial impurities.
Hashem H, et al.
Arabian Journal of Chemistry, 10, S35-S41 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service