T5500
2-Thiobarbituric acid
≥98%
Synonym(s):
4,6-Dihydroxy-2-mercaptopyrimidine, 4,6-Dihydroxypyrimidine-2-thiol, TBA
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Empirical Formula (Hill Notation):
C4H4N2O2S
CAS Number:
Molecular Weight:
144.15
Beilstein:
120663
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Quality Level
Assay
≥98%
drug control
regulated under CDSA - not available from Sigma-Aldrich Canada
mp
245 °C (dec.) (lit.)
SMILES string
O=C1CC(=O)NC(=S)N1
InChI
1S/C4H4N2O2S/c7-2-1-3(8)6-4(9)5-2/h1H2,(H2,5,6,7,8,9)
InChI key
RVBUGGBMJDPOST-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2-Thiobarbituric acid is a molecule made up of heteroatoms: nitrogen, sulphur and oxygen.
Application
2-Thiobarbituric acid has been used to measure malondialdehyde (MDA) in lipid peroxidation assay.
Biochem/physiol Actions
2-Thiobarbituric acid (TBA) is useful to quantitate lipopolysaccharides, carrageenan, and sialic acids. It is also used to detect lipid hydroperoxides and lipid oxidation. It has applications in the field of medicine and biochemistry. TBA may possess the ability to prevent metal corrosion.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Auricularia polytricha aqueous extract supplementation decreases hepatic lipid accumulation and improves antioxidative status in animal model of nonalcoholic fatty liver
Chiu WC, et al.
Biomedicine / [Publiee Pour L'A.A.I.C.I.G.], 4(2) (2014)
The inhibition effect of 2-thiobarbituric acid on the corrosion performance of mild steel in HCl solutions
Kardas G
Materials Science, 41(3), 337-343 (2005)
Evaluation of three 2-thiobarbituric acid methods for the measurement of lipid oxidation in various meats and meat products
Ulu H
Meat Science, 67(4), 683-687 (2004)
Prevention of leukocyte activation by the neutrophil elastase inhibitor, sivelestat, in the hepatic microcirculation after ischemia-reperfusion
Nakano Y, et al.
The Journal of Surgical Research, 155(2), 311-317 (2009)
Leticia R Benetti et al.
European journal of pharmacology, 698(1-3), 463-469 (2012-11-28)
Recent studies show that endogenous hydrogen sulfide (H(2)S) plays an anti-inflammatory role in the pathogenesis of airway inflammation. This study investigated whether exogenous H(2)S may counteract oxidative stress-mediated lung damage in allergic mice. Female BALB/c mice previously sensitized with ovalbumin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service