Skip to Content
Merck
All Photos(1)

Documents

T1700000

Tolbutamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Butyl-3-(4-methylphenylsulfonyl)urea, N-[(Butylamino)carbonyl]-4-methylbenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O3S
CAS Number:
Molecular Weight:
270.35
Beilstein:
1984428
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tolbutamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCNC(=O)NS(=O)(=O)c1ccc(C)cc1

InChI

1S/C12H18N2O3S/c1-3-4-9-13-12(15)14-18(16,17)11-7-5-10(2)6-8-11/h5-8H,3-4,9H2,1-2H3,(H2,13,14,15)

InChI key

JLRGJRBPOGGCBT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tolbutamide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anti-diabetic agent. Metabolized by CYP2C9 (tolbutamide hydroxylase).

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Wang et al.
ACS nano, 9(3), 3159-3168 (2015-03-12)
Cytochromes P450 (CYPs) enzymes are involved in catalyzing the metabolism of various endogenous and exogenous compounds. A rapid analysis of drug metabolism reactions by CYPs is required because they can metabolize 95% of current drugs in drug development and effective
Cheng-Lung Hsu et al.
Oncotarget, 5(8), 2187-2197 (2014-04-20)
Although minoxidil has been used for more than two decades to treat androgenetic alopecia (AGA), an androgen-androgen receptor (AR) pathway-dominant disease, its precise mechanism of action remains elusive. We hypothesized that minoxidil may influence the AR or its downstream signaling.
Paulette B Goforth et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(34), 11405-11415 (2014-08-22)
The adipocyte-derived hormone leptin modulates neural systems appropriately for the status of body energy stores. Leptin inhibits lateral hypothalamic area (LHA) orexin (OX; also known as hypocretin)-producing neurons, which control feeding, activity, and energy expenditure, among other parameters. Our previous
Arian F Baquero et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(30), 9982-9994 (2014-07-25)
Leptin is well known for its role in the regulation of energy homeostasis in adults, a mechanism that at least partially results from the inhibition of the activity of NPY/AgRP/GABA neurons (NAG) in the arcuate nucleus of the hypothalamus (ARH).
Xiwei Zheng et al.
Journal of chromatography. A, 1371, 82-89 (2014-12-03)
Ultrafast affinity extraction and a multi-dimensional affinity system were developed for measuring free drug fractions at therapeutic levels. This approach was used to compare the free fractions and global affinity constants of several sulfonylurea drugs in the presence of normal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service