Skip to Content
Merck
All Photos(1)

Key Documents

G0340000

Glipizide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Cyclohexyl-3-{4-[2-(5-methylpyrazine-2-carboxamido)ethyl]phenylsulfonyl}urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N5O4S
CAS Number:
Molecular Weight:
445.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glipizide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1cnc(cn1)C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3

InChI

1S/C21H27N5O4S/c1-15-13-24-19(14-23-15)20(27)22-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17H,2-6,11-12H2,1H3,(H,22,27)(H2,25,26,28)

InChI key

ZJJXGWJIGJFDTL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glipizide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahra N Mahmoudi et al.
The AAPS journal, 16(4), 685-697 (2014-05-03)
Preparation of amorphous solid dispersions using polymers is a commonly used formulation strategy for enhancing the solubility of poorly water-soluble drugs. However, often a single polymer may not bring about a significant enhancement in solubility or amorphous stability of a
Glipizide: an oral hypoglycemic drug.
G Wensing
The American journal of the medical sciences, 298(1), 69-71 (1989-07-01)
J Ogonowski
Przeglad lekarski, 53(9), 666-667 (1996-01-01)
Glipizide is a second generation of sulphonylurea compound, effective in lowering of blood glucose, well-tolerated as well as safe during long-term treatment. Glipizide is the agent characterized by rapid pharmacological action. It is indicated both in monotherapy and combined treatment
R H Foster et al.
PharmacoEconomics, 18(3), 289-306 (2001-01-09)
Glipizide is a second generation sulphonylurea agent that is available in a Gastrointestinal Therapeutic System (GITS) extended-release formulation. Glipizide GITS provides more stable plasma drug concentrations than the immediate-release formulation and the once-daily regimen may optimise patient compliance. In patients
[The clinical pharmacology of Minidiab (glipizide) (a review of the literature)].
A V Shcherbak
Likars'ka sprava, (1)(1), 28-33 (1993-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service