Merck
Minoxidil may suppress androgen receptor-related functions.
Oncotarget (2014-04-20)
Cheng-Lung Hsu, Jai-Shin Liu, An-Chi Lin, Chih-Hsun Yang, Wen-Hung Chung, Wen-Guey Wu
ABSTRACT
Although minoxidil has been used for more than two decades to treat androgenetic alopecia (AGA), an androgen-androgen receptor (AR) pathway-dominant disease, its precise mechanism of action remains elusive. We hypothesized that minoxidil may influence the AR or its downstream signaling. These tests revealed that minoxidil suppressed AR-related functions, decreasing AR transcriptional activity in reporter assays, reducing expression of AR targets at the protein level, and suppressing AR-positive LNCaP cell growth. Dissecting the underlying mechanisms, we found that minoxidil interfered with AR-peptide, AR-coregulator, and AR N/C-terminal interactions, as well as AR protein stability. Furthermore, a crystallographic analysis using the AR ligand-binding domain (LBD) revealed direct binding of minoxidil to the AR in a minoxidil-AR-LBD co-crystal model, and surface plasmon resonance assays demonstrated that minoxidil directly bound the AR with a K(d) value of 2.6 µM. Minoxidil also suppressed AR-responsive reporter activity and decreased AR protein stability in human hair dermal papilla cells. The current findings provide evidence that minoxidil could be used to treat both cancer and age-related disease, and open a new avenue for applications of minoxidil in treating androgen-AR pathway-related diseases.
Sigma-Aldrich
Minoxidil, ≥99% (TLC)
Pricing and availability is not currently available.
Bicalutamide, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Minoxidil, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Tolbutamide, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Pricing and availability is not currently available.
Tolbutamide, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.