Skip to Content
Merck
All Photos(1)

Documents

SMB01337

Sigma-Aldrich

DIMBOA

Synonym(s):

(±)-DIMBOA, 2,4-Dihydroxy-7-methoxy-1,4-benzoxazin-3(4H)-one, 2,4-Dihydroxy-7-methoxy-1,4-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-2H-1,4-benzoxazin-3(4H)-one, 2,4-Dihydroxy-7-methoxy-2H-benzo[b][1,4]oxazin-3(4H)-one, 2,4-dihydroxy-7-methoxy- (6CI, 7CI, 8CI, 9CI, ACI), 2H-1,4-Benzoxazin-3(4H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO5
Molecular Weight:
211.17
MDL number:
NACRES:
NA.77

Assay

≥92% (HPLC)

Quality Level

form

solid

mp

150—155 °C

storage temp.

2-8°C

General description

DIMBOA is a powerful antibiotic present in maize and wheat. It was first identified in maize in 1962 as a saccharin-like ″sweet substance in corn″. DIMBOA is stored as an inactive precursor, DIMBOA-glucoside, which is activated by glucosidases in response to insect feeding. In maize, DIMBOA functions as natural defense against damaging insect pests and pathogens, including fungi and bacteria. DIMBOA level varies among individual plants, higher concentrations are typically found in young seedlings and lower concentrations in adult maize plants.

Application

Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cuiping Zhang et al.
The New phytologist, 229(4), 2273-2287 (2020-10-01)
2,4-Dihydroxy-7-methoxy-1,4-benzoxazin-3-one (DIMBOA) and DIMBOA-glucoside (DIMBOA-Glc) are anti-insect benzoxazinoids in maize, yet very little information is known about how they are regulated. Reverse genetics, kinase activity analysis, phytohormone and DIMBOA/DIMBOA-Glc quantification, bioassays and transcriptome analysis were employed to study the function
A M Giuffrida Stella et al.
Gerontology, 33(3-4), 136-148 (1987-01-01)
During aging there are several structural, functional and biochemical alterations, including changes in macromolecular composition and turnover. Regulation of gene expression, DNA and RNA synthesis, total poly(A)+ and poly(A)- RNA contents, qualitative and quantitative changes of synaptosomal plasma membrane proteins
Karina Lucas Silva-Brandão et al.
Pest management science, 77(1), 518-526 (2020-08-21)
2,4-dihydroxy-7-methoxy-1,4-benzoxazin-3-one (DIMBOA), the main benzoxazinoid found in corn, elicits variable larval responses from different pest moths. For the widespread and highly polyphagous Spodoptera frugiperda (Lepidoptera: Noctuidae), the fall-armyworm (FAW), DIMBOA acts as a feeding stimulant and improves larval growth at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service