Skip to Content
Merck
All Photos(2)

Key Documents

T1438

Millipore

Todd Hewitt broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

7.8±0.2 (25 °C)

application(s)

clinical testing
food and beverages

microbiology

storage temp.

2-25°C

suitability

nonselective for Streptococcus spp.

General description

Todd Hewitt Broth is used for cultivation of haemolytic streptococci for different serological tests. This medium has been recommended as an alternative type in epidemiologic studies of group A streptococci as well as pathogenic microorganisms. Todd Hewitt Broth medium is very nutritive by the presence of peptic digest of animal tissue and beef heart infusion. Dextrose stimulates haemolysin production. The medium is well buffered by sodium phosphate and sodium carbonate to neutralize the acid produced during dextrose fermentation. This restricts destruction of antigenic streptococcal haemolysin. It is also found that sodium phosphate has a stimulating effect on the pneumococcal growth. Todd Hewitt Broth can be employed as an alternative to serum broth or horse flesh digest broth for the cultivation of streptococci prior to serological typing.

Application

Recommended for the cultivation of group A hemolytic Streptococci used for serological studies.
Todd Hewitt broth has been used for the culture of various microorganisms, such as Bacillus subtilis and Streptococcus pneumoniae.

Components

Components (g/L):
Beef Heart Infusion 500.00
Peptic Digest of Animal Tissue 20.00
Dextrose 2.00
Sodium Chloride 2.00
Sodium Phosphate 0.40
Sodium Carbonate 2.50

Preparation Note

Suspend 37 g of Todd Hewitt Broth in 1000 ml of distilled water. Mix well and dispense as desired. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alcamo's Laboratory Fundamentals of Microbiology (2010)
Finegold, S.M., et al.
Bailey and Scott's Diagnostic Microbiology (1982)
Handbook of Microbiological Media Third Edition (2004)
The pore-forming protein gasdermin D regulates interleukin-1 secretion from living macrophages
Evavold CL, et al.
Immunity, 48(1), 35-44 (2018)
Assessing membrane fluidity and visualizing fluid membrane domains in bacteria using fluorescent membrane dyes
Wenzel M, et al.
Bio-protocol (2018)

Articles

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service