Skip to Content
Merck
All Photos(2)

Key Documents

Y1625

Sigma-Aldrich

Yeast Extract

for use in microbial growth medium

Synonym(s):

Select Yeast Extract, microbial growth media, microbial growth medium

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
eCl@ss:
42040000
NACRES:
NA.85

grade

Molecular Biology
for molecular biology

Quality Level

sterility

non-sterile

form

powder

technique(s)

microbiological culture: suitable using





application(s)

agriculture

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

SMILES string

OC(=O)CC1c2c3c(ccc2c4c1cccc4)cccc3

InChI

1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)

InChI key

GQNBDGXKDJSVGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Yeast extract is produced by various yeast species mainly by Saccharomyces cerevisiae or baker’s yeast. In addition to its use as a supplement in culture media for microbial research and biotechnology, yeast extract is also used as a component of the complex media for industrial fermentations.It also serves as a nutritional source for various microbial media.

Application

For general bacteriological use with a variety of microorganisms.
Yeast Extract has been used as a component of:
  • complex media for culturing E. coli clones for shake flask experiments
  • culture media for culturing bacterial species Gluconacetobacter xylinus
  • yeast extract beef (YEB) media for the transformation of Agrobacterium
Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.

Other Notes

Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco R Binelli et al.
Biomaterials advances, 141, 213095-213095 (2022-09-06)
Bacterial cellulose is an attractive resource for the manufacturing of sustainable materials, but it is usually challenging to shape it into elaborate three-dimensional structures. Here, we report a manufacturing platform for the creation of complex-shaped cellulose objects by printing inks
Luciana M Katiki et al.
Veterinary parasitology, 182(2-4), 264-268 (2011-06-18)
The most challenging obstacles to testing products for their anthelmintic activity are: (1) establishing a suitable nematode in vitro assay that can evaluate potential product use against a parasitic nematode of interest and (2) preparation of extracts that can be
Jinyou Zhang et al.
Biotechnology and bioengineering, 82(6), 640-652 (2003-04-04)
Yeast extract (YE) is commonly used as a key component in the complex media for industrial fermentations. However, the lot-to-lot variation of this raw material frequently requires extensive "use testing" of many lots to identify only the few that support
Omid Zarei et al.
Iranian journal of pharmaceutical research : IJPR, 15(4), 907-913 (2017-03-01)
Yeasts, especially Saccharomyces cerevisiae, are one of the oldest organisms with broad spectrum of applications, owing to their unique genetics and physiology. Yeast extract, i.e. the product of yeast cells, is extensively used as nutritional resource in bacterial culture media.
Pascal Chatonnet et al.
Journal of agricultural and food chemistry, 58(23), 12481-12490 (2010-11-10)
This study identifies a previously isolated bacterium as Rhizobium excellensis, a new species of proteobacteria able to form a large quantity of 2-methoxy-3,5-dimethylpyrazine (MDMP). R. excellensis actively synthesizes MDMP from L-alanine and L-leucine and, to a lesser extent, from L-phenylalanine

Protocols

TE Buffer; Elution Buffer; 10x Ligation Buffer; 0.5 M PIPES Buffer; Inoue Transformation Buffer

TE Buffer; Elution Buffer; 10x Ligation Buffer; 0.5 M PIPES Buffer; Inoue Transformation Buffer

TE Buffer; Elution Buffer; 10x Ligation Buffer; 0.5 M PIPES Buffer; Inoue Transformation Buffer

TE Buffer; Elution Buffer; 10x Ligation Buffer; 0.5 M PIPES Buffer; Inoue Transformation Buffer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service