Skip to Content
Merck
All Photos(1)

Key Documents

C7642

Sigma-Aldrich

Concanavalin A from Canavalia ensiformis (Jack bean)

FITC conjugate, Type IV, lyophilized powder

Synonym(s):

ConA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Canavalia ensiformis

Quality Level

conjugate

FITC conjugate

type

Type IV

form

lyophilized powder

composition

Protein, 5-15% biuret

extent of labeling

3-6 mol FITC per mol protein

impurities

Carbohydrate, essentially free

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

For the fluorescent detection of glycoproteins containing α−D-mannose or α−D-glucose

Biochem/physiol Actions

Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Ca2+ and Mn2+ ions are required for activity. Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer; above pH 7.0 higher aggregates are formed. Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6.

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing phosphate buffer salts and NaCl

Analysis Note

Where reported, agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, containing Ca2+ and Mn2+ of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25 °C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Liu et al.
Molecular biology and evolution, 37(8), 2300-2308 (2020-04-04)
Fisher's fundamental theorem of natural selection predicts no additive variance of fitness in a natural population. Consistently, studies in a variety of wild populations show virtually no narrow-sense heritability (h2) for traits important to fitness. However, counterexamples are occasionally reported
Jianbin Guo et al.
International journal of nanomedicine, 16, 7249-7268 (2021-11-06)
With increases in implant infections, the search for antibacterial and biofilm coatings has become a new interest for orthopaedists and dentists. In recent years, graphene oxide (GO) has been extensively studied for its superior antibacterial properties. However, most of these
Sanda Ljubicic et al.
Biology of the cell, 112(12), 427-438 (2020-08-29)
Cell-cell or cell-substrate interactions are lost when cells are dissociated in culture, or during pathophysiological breakdowns, therefore impairing their structure and polarity, and affecting their function. We show that single rat β-cells, cultured under non-adhesive conditions, form intracytoplasmic vacuoles increasing
Uma Shankar Sagaram et al.
Microbiology (Reading, England), 153(Pt 9), 2850-2861 (2007-09-05)
Fusarium verticillioides is an important pathogen of maize that causes ear rot and produces the mycotoxins known as fumonisins. To date, knowledge of pathogenicity and the regulation of fumonisin biosynthesis in F. verticillioides is limited. Here, the molecular characterization of
Jianhong Zhou et al.
Biochemical pharmacology, 180, 114108-114108 (2020-06-23)
Noninfectious (autoimmune and immune-mediated) uveitis is an ocular inflammatory disease which can lead to blindness in severe cases. Due to the potential side effects of first-line drugs for clinical uveitis, novel drugs and targets against uveitis are still urgently needed.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service