Skip to Content
Merck
All Photos(1)

Documents

C2219000

Citric acid monohydrate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2 · H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
4018641
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

citric acid

manufacturer/tradename

EDQM

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(CC(O)(C(O)=O)CC(O)=O)=O.O

InChI

1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2

InChI key

YASYEJJMZJALEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Citric acid monohydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

345.0 °F - closed cup

Flash Point(C)

173.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuhal Guvenalp et al.
Pharmaceutical biology, 53(6), 888-896 (2014-11-29)
Mentha L. (Labiatae) species (mint) with their flavoring properties have been used in food industries for centuries. Besides they have a great importance in drug development and medicinal applications due to various bioactive compounds of several members of the genus.
Katja Sundström et al.
Endocrinology, 155(12), 4798-4807 (2014-09-23)
The growth-promoting effect of combined therapy with GH and IGF-I in normal rats is not known. We therefore investigated the efficacy of treatment with recombinant human (rh)GH and/or rhIGF-I on longitudinal bone growth and bone mass in intact, prepubertal, female
Anne Marie V Schou-Pedersen et al.
Journal of pharmaceutical and biomedical analysis, 107, 333-340 (2015-02-04)
Concentrated solutions containing 6-aminocaproic acid and the excipients citric acid and sorbitol have been studied at temperatures of 50°C, 60°C, 70°C and 80°C as well as at 20°C. It has previously been reported that the commonly employed citric acid is
Anne R Tinning et al.
American journal of physiology. Renal physiology, 307(11), F1215-F1226 (2014-10-24)
Both the processing and release of secretory granules involve water movement across granule membranes. It was hypothesized that the water channel aquaporin (AQP)1 directly contributes to the recruitment of renin-positive cells in the afferent arteriole. AQP1(-/-) and AQP1(+/+) mice were
Leila Bagheri et al.
Journal of microencapsulation, 31(7), 636-643 (2014-04-29)
Whey proteins were inter-connected either by the enzyme transglutaminase or citric acid and then desolvated with ethanol to generate particles. Both samples comprised of sub-micron (>300 nm) and nano-scaled (~100 nm) particles based on the hydrodynamic size measurements. Enzyme-induced cross-linking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service