Skip to Content
Merck
All Photos(4)

Key Documents

C0759

Sigma-Aldrich

Citric acid

99%

Synonym(s):

2-Hydroxy-1,2,3-propanetricarboxylic acid, 2-Hydroxypropan-1,2,3-tricarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021801
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

≤ 0.1 hPa ( at 25 °C)

Quality Level

Assay

99%

form

crystals

expl. lim.

8 %, 65 °F

concentration

≤100%

pH

ca.1,7-at 100 g/l-at 20 °C

pKa (25 °C)

(1) 3.13, (2) 4.76, (3) 6.40

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

bp

200 °C/at 1.013 hPa (decomposition)

mp

153-159 °C (lit.)

solubility

water: soluble 383 g/L at 25 °C

density

1,67 g/cm3 at at 20 °C

functional group

carboxylic acid
hydroxyl

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Citric acid is a natural component widely usedin many industrial applications such as chelation, buffering, pH adjustment,and derivatization. It can also be used as a reagent in condensation, acidcatalysis, cycloaddition, deprotection, and decarboxylation reactions.

Application

Citric acid can be used as a reactant to synthesize:
  • Trialkyl citrates via sulfuric acid-catalyzed esterification with aliphatic alcohols C2-C5.
  • Biodegradable thermoset polymer via catalyst-free polycondensation reaction.
It can also be used as a catalyst to synthesize amido alkyl naphthols by reacting 2-naphthols, aldehydes, and amides/ureas.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method for decalcification with citric acid.
Foschini MP and Muzzi L.
Biotechnic & Histochemistry, 68(1), 42-45 (1993)
Wheat straw hemicellulose films as affected by citric acid.
Azeredo HMC, et al.
Food Hydrocolloids, 50 (2015)
Determination of citric acid in fats.
Ohlson R, et al.
Journal of the American Oil Chemists' Society, 45(6), 475-476 (1968)
Factors affecting the successful cryopreservation of honey bee (Apis mellifera) spermatozoa.
Hopkins BK and Herr C.
Apidologie, 41(5), 548-556 (2010)
Assay of Ornithine Decarboxylase and Spermidine/Spermine N1-acetyltransferase Activities.
Hyvonen MT, et al.
Bio-protocol, 4(22) (2014)

Articles

Citrate is one of many cell culture components that affects the performance of serum-free, protein-free and animal protein-free cell culture systems used for biomanufacturing heterologous proteins such as monoclonal antibodies.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service