Skip to Content
Merck
All Photos(2)

Documents

B0126

Sigma-Aldrich

Bromophenol Blue

Powder

Synonym(s):

3′,3′′,5′,5′′-Tetrabromophenolsulfonephthalein, Bromophenol Blue (BPB), Bromphenol Blue Sultone Form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H10Br4O5S
CAS Number:
Molecular Weight:
669.96
Beilstein:
61698
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Bromophenol Blue, titration: suitable

Agency

suitable for SM 2310
suitable for SM 2320

Quality Level

form

powder

technique(s)

titration: suitable

color

faint beige to very dark beige
faint orange to very dark orange

visual transition interval

3.0-4.6, yellow-green to blue

mp

273 °C

solubility

methanol: 10 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(Br)cc(cc1Br)C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c(O)c(Br)c4

InChI

1S/C19H10Br4O5S/c20-12-5-9(6-13(21)17(12)24)19(10-7-14(22)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19/h1-8,24-25H

InChI key

UDSAIICHUKSCKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromophenol blue is present as a sulfonated hydroxyquinone in acidic aqueous solutions. It is also known as tetrabromophenolsulfonphthalein. Bromophenol Blue is a pH indicator and a dye appears a strong blue color. It has a slightly negative charge and will migrate in the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.

Application

Bromophenol Blue is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis. It has been used in the preparation of protein samples for western blotting analysis. Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.It has also been known for its use as a vital stain to probe the blood-brain barrier.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

In vitro Reconstitution Assay of miRNA Biogenesis by Arabidopsis DCL1.
Wang T, et al.
Bio-protocol, 5, pii: e1454-pii: e1454 (2015)
Effective elimination of chronic lymphocytic leukemia cells in the stromal microenvironment by a novel drug combination strategy using redox-mediated mechanisms.
Zhang W, et al.
Molecular Medicine Reports, 12, 7374-7388 (2015)
Ibrahim Alkatout et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 28(1), 69-79 (2014-06-21)
The prognosis of pancreatic neuroendocrine tumors is related to size, histology and proliferation rate. However, this stratification needs to be refined further. We conducted a proteome study on insulinomas, a well-defined pancreatic neuroendocrine tumor entity, in order to identify proteins
Dana Westphal et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(39), E4076-E4085 (2014-09-18)
The pivotal step on the mitochondrial pathway to apoptosis is permeabilization of the mitochondrial outer membrane (MOM) by oligomers of the B-cell lymphoma-2 (Bcl-2) family members Bak or Bax. However, how they disrupt MOM integrity is unknown. A longstanding model
Stephanie L Smith-Roe et al.
Oncotarget, 6(2), 732-745 (2014-12-30)
SWI/SNF complexes utilize BRG1 (also known as SMARCA4) or BRM (also known as SMARCA2) as alternative catalytic subunits with ATPase activity to remodel chromatin. These chromatin-remodeling complexes are required for mammalian development and are mutated in ~20% of all human

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service