Skip to Content
Merck
All Photos(3)

Documents

B8026

Sigma-Aldrich

Bromophenol Blue sodium salt

For molecular biology, powder

Synonym(s):

3′,3′′,5′,5′′-Tetrabromophenolsulfophthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H9Br4NaO5S
CAS Number:
Molecular Weight:
691.94
Beilstein:
6995701
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Bromophenol Blue sodium salt, for molecular biology, suitable for electrophoresis

grade

for molecular biology

Quality Level

form

powder

technique(s)

electrophoresis: suitable

visual transition interval

3.0-4.6, yellow to blue

ε (extinction coefficient)

≥150000 at 597-603 nm in H2O
≥17000 at 250-256 nm in H2O
≥23000 at 306-312 nm in H2O

suitability

suitable for (Suitable For Use as a Tracking Dye For Electrophoresis in Alkaline and Neutral Buffers)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Oc1c(Br)cc(cc1Br)C(=C2/C=C(Br)C(=O)C(Br)=C2)\c3ccccc3S([O-])(=O)=O

InChI

1S/C19H10Br4O5S.Na/c20-12-5-9(6-13(21)18(12)24)17(10-7-14(22)19(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28;/h1-8,24H,(H,26,27,28);/q;+1/p-1

InChI key

TWKDJMRHSLWRFI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Bromophenol blue (BroB), an arylmethane color marker dye has a molecular mass of 670 Da.

Application

Bromophenol Blue sodium salt has been used in the buffer solution to differentiate dead and living cells. It has also been used as a constituent in gel loading buffer.
A tracking dye for nucleic acid or protein electrophoresis in agarose or polyacrylamide gels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of corticotherapy on proteomics of endometrial fluid from mares susceptible to persistent postbreeding endometritis.
Wolf CA, et al.
Theriogenology, 77, 1351-1359 (2012)
The expression of ERa, OTR, cPLA(2), COX-2, and PPAR? in the cervix of the ewe during the estrous cycle.
Falchi L and Scaramuzzi RJ
Theriogenology, 79, 40-47 (2013)
Rapid transfer of DNA from agarose gels to nylon membranes.
Reed KC and Mann DA
Nucleic Acids Research, 13.20, 7207-7221 (1985)
A dielectrophoretic continuous flow sorter using integrated microelectrodes coupled to a channel constriction
Salomon S, et al.
Electrophoresis, 32(12), 1508-1514 (2011)
Purification of human plasma fibronectin using immobilized gelatin and Arg affinity chromatography
Speziale P, et al.
Nature Protocols, 3(3), 525-525 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service