Skip to Content
Merck
All Photos(3)

Documents

114391

Sigma-Aldrich

Bromophenol Blue

ACS reagent

Synonym(s):

3′,3″,5′,5″-Tetrabromophenolsulfonphthalein, Bromphenol Blue Sultone Form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H10Br4O5S
CAS Number:
Molecular Weight:
669.96
Beilstein:
61698
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

powder

technique(s)

titration: suitable

visual transition interval

3.0-4.6, yellow to blue

mp

273 °C (lit.)

solubility

ethanol: 0.1%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(Br)cc(cc1Br)C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c(O)c(Br)c4

InChI

1S/C19H10Br4O5S/c20-12-5-9(6-13(21)17(12)24)19(10-7-14(22)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19/h1-8,24-25H

InChI key

UDSAIICHUKSCKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromophenol blue is present as a sulfonated hydroxyquinone in acidic aqueous solutions. It is also known as tetrabromophenolsulfonphthalein.Bromophenol Blue is a pH indicator and a dye appearing as a strong blue color. It has a slight negative charge and will migrate the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.

Application

Bromphenol Blue is used as a tracking dye in DNA, RNA (agarose), and protein (polyacrylamide) gel electrophoresis. It has been used in the preparation of protein samples for western blotting analysis. Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels. It has also been known for its use as a vital stain to probe the blood-brain barrier.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahim Alkatout et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 28(1), 69-79 (2014-06-21)
The prognosis of pancreatic neuroendocrine tumors is related to size, histology and proliferation rate. However, this stratification needs to be refined further. We conducted a proteome study on insulinomas, a well-defined pancreatic neuroendocrine tumor entity, in order to identify proteins
Stephanie L Smith-Roe et al.
Oncotarget, 6(2), 732-745 (2014-12-30)
SWI/SNF complexes utilize BRG1 (also known as SMARCA4) or BRM (also known as SMARCA2) as alternative catalytic subunits with ATPase activity to remodel chromatin. These chromatin-remodeling complexes are required for mammalian development and are mutated in ~20% of all human
Dana Westphal et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(39), E4076-E4085 (2014-09-18)
The pivotal step on the mitochondrial pathway to apoptosis is permeabilization of the mitochondrial outer membrane (MOM) by oligomers of the B-cell lymphoma-2 (Bcl-2) family members Bak or Bax. However, how they disrupt MOM integrity is unknown. A longstanding model
Ákos Kenéz et al.
PloS one, 10(5), e0127208-e0127208 (2015-05-16)
Dairy cows mobilize large amounts of body fat during early lactation to overcome negative energy balance which typically arises in this period. As an adaptation process, adipose tissues of cows undergo extensive remodeling during late pregnancy and early lactation. The
Sing Ling Chia et al.
Small (Weinheim an der Bergstrasse, Germany), 11(6), 702-712 (2014-10-22)
Our current mechanistic understanding on the effects of engineered nanoparticles (NPs) on cellular physiology is derived mainly from 2D cell culture studies. However, conventional monolayer cell culture may not accurately model the mass transfer gradient that is expected in 3D

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service