Skip to Content
Merck
All Photos(1)

Documents

97865

Supelco

1,3-Diethylbenzene

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903394
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.496

density

0.864 g/mL at 20 °C (lit.)

SMILES string

CCc1cccc(CC)c1

InChI

1S/C10H14/c1-3-9-6-5-7-10(4-2)8-9/h5-8H,3-4H2,1-2H3

InChI key

AFZZYIJIWUTJFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Diethylbenzenes are aromatic hydrocarbons that are classified under the class of volatile organic compounds (VOCs).

Application

1,3-Diethylbenzene may be used as an analytical reference standard for the analysis of 1,3-diethylbenzene in:
  • White spirit samples and Chinese crude oils by comprehensive two-dimensional gas chromatography coupled to time-of-flight mass spectrometry (GC x GC TOFMS) operating on electron impact (EI) mode.
  • Outdoor air samples by sampling on adsorbent tubes followed by determination using thermal desorption (TD) combined with GC-MS.
  • Virgin olive oils by GC in combination with ion trap EI-MS.
  • Atmospheric air samples by GC x GC TOFMS and field olfactometry techniques.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of odour concentration by TD-GC x GC-TOF-MS and field olfactometry techniques.
Bylinski H, et al.
Monatshefte fur Chemie / Chemical Monthly, 148(9), 1651-1659 (2017)
Prospective application of melanized fungi for the biofiltration of indoor air in closed bioregenerative systems.
Prenafeta-Boldu FX, et al.
Journal of Hazardous Materials, 361, 1-9 (2019)
Kanji Yamasaki et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(8), 2635-2642 (2012-05-31)
The purpose of this study was to investigate the endocrine-mediated effects of the benzene-related compounds with reference to Organization for Economic Co-operation and Development (OECD) Test Guideline No. 407. Rats were orally gavaged with 0, 10, 50, and 250 mg/kg/day
Towards chemical profiling of ignitable liquids with comprehensive two-dimensional gas chromatography: Exploring forensic application to neat white spirits.
Sampat AAS, et al.
Forensic Science International, 267, 183-195 (2016)
New approach to resolve the humidity problem in VOC determination in outdoor air samples using solid adsorbent tubes followed by TD-GC-MS.
Maceira A, et al.
The Science of the Total Environment, 599, 1718-1727 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service