CRMUMPGO
Diesel
Multi-Parameter Certified Reference Material
Synonym(s):
Diesel MPCRMs
About This Item
Recommended Products
grade
certified reference material
form
liquid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Qualified for viscosity, density, flash point, cloud point, cold filter plugging point (CFPP) and distillation using ASTM / IP and EN test method protocols
- Low levels of uncertainty ensures maximum accuracy of data
- Fully traceable to international standards
- 18-month shelf life
- 500 mL inert glass packaging with tamper-evident cap provides absolute assurance of sample integrity
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
134.6 °F
Flash Point(C)
57 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service