Skip to Content
Merck
All Photos(2)

Key Documents

414565

Sigma-Aldrich

Divinylbenzene

technical grade, 80%

Synonym(s):

Diethenylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH=CH2)2
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

technical grade

vapor density

4.5 (vs air)

vapor pressure

0.9 mmHg ( 30 °C)

Assay

80%

form

liquid

autoignition temp.

878 °F

contains

1000 ppm p-tert-butylcatechol as inhibitor

expl. lim.

0.7 %, 75 °F
6.5 %, 130 °F

refractive index

n20/D 1.561 (lit.)

bp

195 °C (lit.)

density

0.914 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=Cc1ccc(C=C)cc1

InChI

1S/C10H10/c1-3-9-7-5-6-8-10(9)4-2/h3-8H,1-2H2

InChI key

MYRTYDVEIRVNKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Divinylbenzene (DVB) consists of a combination of meta and para isomers of DVB and ethylvinylbenzene. It can be used as a monomer for the preparation of linear polydivinylbenzene by anionic polymerization.[1][2]
Divinylbenzene (DVB) is a bis unsaturated monomer. DVB along with styrene as a co monomer is used in producing cross linked polymers. Several examples have reported. Polymerization with high concentrations of DVB produces popcorn polymers.

Application

DVB can be co-polymerized with sulfur due to its high reactivity. It can be used as a cathode material in lithium-sulfur batteries.[3] It can also be used as a cross-linker that increases the rigidity and reduces the template leakage of the molecular imprinted polymers (MIPs).[4] DVB can be polymerized to form poly(divinylbenzene) that can be carbonized to fabricate activated carbon monoliths.[5]
DVB is used in polymeric core shell nanoparticles[6] and as a crosslinking agent.[7] DVB may be used as a monomer for the synthesis of highly crosslinked macroporous styrene/DVB polymers.[8] It may also be used to synthesize monodispersed poly-DVB microspheres.[9]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization, and application of novel polymeric nanoparticles.
Wang X, et al.
Macromolecules, 40(3), 499-508 (2007)
Inverse vulcanization of sulfur with divinylbenzene: Stable and easy processable cathode material for lithium-sulfur batteries.
Gomez I, et al.
Journal of Power Sources, 329(4), 72-78 (2016)
Sensors based on surface plasmon resonance in a plastic optical fiber for the detection of trinitrotoluene.
Cennamo N, et al.
Sensors and Actuators B, Chemical, 188, 221-226 (2013)
Fabrication of activated carbons with well-defined macropores derived from sulfonated poly (divinylbenzene) networks.
Hasegawa, et al.
Carbon, 48(6), 1757-1766 (2010)
Porous monodisperse poly(divinylbenzene) microspheres by precipitation polymerization
Li WH, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 36, 1543-1551 null

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service