Skip to Content
Merck
All Photos(2)

Documents

45935

Supelco

1,2,3-Trimethylbenzene

analytical standard

Synonym(s):

Hemellitol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
1903410
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39011402
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.15 (vs air)

vapor pressure

3.4 mmHg ( 37.7 °C)

autoignition temp.

878 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.513 (lit.)

bp

175-176 °C (lit.)

mp

−25 °C (lit.)

density

0.894 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1cccc(C)c1C

InChI

1S/C9H12/c1-7-5-4-6-8(2)9(7)3/h4-6H,1-3H3

InChI key

FYGHSUNMUKGBRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Tomas et al.
International journal of occupational medicine and environmental health, 12(1), 73-84 (1999-06-09)
In a series of acute experiments on rats the potential of toluene, mesitylene, hemimellitene and pseudocumene to affect the CNS function was assessed following an analysis of spontaneous and evoked hippocampal and cortical activity. The electrophysiological examinations were performed on
Y Tsujimoto et al.
Chemosphere, 39(5), 725-730 (1999-08-17)
The structure was investigated of the mercapturic acid excreted in urine of rats after the i.p. administration of 1,2,3-trimethylbenzene. Of the two regioisomeric mercapturic acids, i.e. N-acetyl-S-(2,3-dimethylbenzyl)-L-cysteine and N-acetyl-S-(2,6-dimethyl-benzyl)-L-cysteine, only the former was isolated by preparative HPLC and identified, by
Z Korsak et al.
International journal of occupational medicine and environmental health, 13(3), 223-232 (2000-12-08)
Toxic effects of exposure to 1,2,3-trimethylbenzene (hemimellitene) in the condition of subchronic inhalation experiment were examined. Rats were exposed to vapours of hemimellitene at concentrations of 123 mg/m3, 492 mg/m3 and 1230 mg/m3, 6 h/day, 5 days/week for 3 months.
Application of gas chromatography in conjunction with enrichment of samples on active carbon to determine low concentrations of organic compounds in air. Part I. Aromatic hydrocarbons.
H Swedrowska et al.
Bulletin of the Institute of Maritime and Tropical Medicine in Gdynia, 37(1-2), 103-112 (1986-01-01)
Z Korsak et al.
International journal of occupational medicine and environmental health, 9(4), 341-349 (1996-01-01)
Neurotoxic effects of trimethylbenzene isomers (pseudocumene, mesitylene and hemimellitene) in male rats were investigated in conditions of acute and subchronic inhalation exposure. Rotarod performance and pain sensitivity behaviour were tested in rats exposed to trimethylbenzenes at concentrations of 250-2,000 ppm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service