Skip to Content
Merck
All Photos(2)

Key Documents

75614

Supelco

2-Palmitoylglycerol

analytical standard

Synonym(s):

β-Monopalmitin, 2-Hexadecanoylglycerol, 2-Monohexadecanoylglycerol, 2-Monopalmitin, 2-Monopalmitoylglycerol, Hexadecanoic acid 2-hydroxy-1-(hydroxymethyl)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H38O4
CAS Number:
Molecular Weight:
330.50
Beilstein:
1796692
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

Quality Level

grade

analytical standard

Assay

≥95% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

ester

storage temp.

−70°C

SMILES string

O=C(CCCCCCCCCCCCCCC)OC(CO)CO

InChI

1S/C19H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-18(16-20)17-21/h18,20-21H,2-17H2,1H3

InChI key

BBNYCLAREVXOSG-UHFFFAOYSA-N

Application

2-Palmitoylglycerol may be used as a reference standard for the separation and determination of the analyte in partial glyceride mixtures using thin layer chromatography with flame ionization detection (TLC-FID).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of isomeric glycerides, free fatty acids and triglycerides by thin layer chromatography-flame ionization detector system.
Tanaka M, et al.
Methods, 15(10), 872-875 (1980)
Elizabeth J Judge et al.
Rapid communications in mass spectrometry : RCM, 31(20), 1690-1698 (2017-08-10)
Positional analysis of intact triglycerides could provide greater insights into the link between fatty acid position and lipotoxic diseases. However, this methodology has been impeded by lack of commercial availability of positionally pure triglycerides. This work reports on a strategy
Biswajit Khatua et al.
Science advances, 7(5) (2021-01-31)
Obesity sometimes seems protective in disease. This obesity paradox is predominantly described in reports from the Western Hemisphere during acute illnesses. Since adipose triglyceride composition corresponds to long-term dietary patterns, we performed a meta-analysis modeling the effect of obesity on
Caroline Turcotte et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(3), 4253-4265 (2020-02-06)
The endocannabinoid (eCB) 2-arachidonoyl-gycerol (2-AG) modulates immune responses by activating cannabinoid receptors or through its multiple metabolites, notably eicosanoids. Thus, 2-AG hydrolysis inhibition might represent an interesting anti-inflammatory strategy that would simultaneously increase the levels of 2-AG and decrease those
Avery L McIntosh et al.
Journal of lipid research, 59(4), 646-657 (2018-02-08)
Phytocannabinoids, such as Δ9-tetrahydrocannabinol (THC), bind and activate cannabinoid (CB) receptors, thereby "piggy-backing" on the same pathway's endogenous endocannabinoids (ECs). The recent discovery that liver fatty acid binding protein-1 (FABP1) is the major cytosolic "chaperone" protein with high affinity for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service