Skip to Content
Merck
All Photos(4)

Key Documents

D2135

Sigma-Aldrich

1,2-Dipalmitoyl-rac-glycerol

≥99%

Synonym(s):

(±)-1,2-Dipalmitin, 1,2-Dihexadecanoyl-rac-glycerol, rac-Glycerol 1,2-dipalmitate, DL-1,2-Dipalmitin, DG(16:0/16:0/0:0)[rac], Hexadecanoic acid 1,1′-[1-(hydroxymethyl)-1,2-ethanediyl] ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H68O5
CAS Number:
Molecular Weight:
568.91
Beilstein:
1809777
EC Number:
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C35H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-32-33(31-36)40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33,36H,3-32H2,1-2H3

InChI key

JEJLGIQLPYYGEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacinta L Watkins et al.
The Plant cell, 31(12), 3092-3112 (2019-10-03)
Xanthophylls are a class of carotenoids that are important micronutrients for humans. They are often found esterified with fatty acids in fruits, vegetables, and certain grains, including bread wheat (Triticum aestivum). Esterification promotes the sequestration and accumulation of carotenoids, thereby
Tomoyuki Ozawa et al.
Journal of mass spectrometry : JMS, 50(11), 1264-1269 (2015-10-28)
Matrix-assisted laser desorption/ionisation (MALDI) imaging mass spectrometry (IMS) allows for the simultaneous detection and imaging of several molecules in brain tissue. However, the detection of glycerolipids such as diacylglycerol (DAG) and triacylglycerol (TAG) in brain tissues is hindered in MALDI-IMS
Vincent O Oninla et al.
Journal of lipid research, 55(12), 2606-2619 (2014-10-24)
During endocytosis, membrane components move to intraluminal vesicles of the endolysosomal compartment for digestion. At the late endosomes, cholesterol is sorted out mainly by two sterol-binding proteins, Niemann-Pick protein type C (NPC)1 and NPC2. To study the NPC2-mediated intervesicular cholesterol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service