Skip to Content
Merck
All Photos(1)

Documents

18109

Supelco

Hexanal

analytical standard

Synonym(s):

Aldehyde C6, Caproaldehyde, Hexyl aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CHO
CAS Number:
Molecular Weight:
100.16
Beilstein:
506198
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

10 mmHg ( 20 °C)

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

refractive index

n20/D 1.4035 (lit.)

bp

130-131 °C (lit.)

density

0.815 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCC=O

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h6H,2-5H2,1H3

InChI key

JARKCYVAAOWBJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexanal is an oxidation product of linoleic acid and is a dominant volatile aldehyde. It could work as an indicator of lipid oxidation for cooked meat and meat flavor deterioration.

Application

It was used as standard for measuring lipid peroxidation in rat liver samples using headspace GC analysis.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rapid headspace gas chromatography of hexanal as a measure of lipid peroxidation in biological samples.
Frankel EN, Hu ML, Tappel AL
Lipids, 24(11), 976-981 (1989)
Hexanal as an indicator of meat flavor deterioration.
Shahidi, Fereidoon, and RONALD B. PEGG.
Journal of Food Lipids, 1.3, 177-186 (1994)
Jie Li et al.
Clinica chimica acta; international journal of clinical chemistry, 436, 59-67 (2014-05-13)
Breast cancer (BC) remains the most commonly diagnosed malignancy in women. We investigated 4 straight aldehydes in the exhaled breath as potential early BC diagnostic biomarkers. End-tailed breath were collected by Bio-VOC® sampler and assayed by gas chromatography-mass spectrometry. Kruskal-Wallis
S Böttcher et al.
Food chemistry, 169, 492-498 (2014-09-23)
The aim of the present study was to evaluate the off-flavour masking potential of pea dextrin (PD) in emulsions rich in ω-3 and ω-6-fatty acids in comparison with maltodextrin (MD) and 2-hydroxypropyl-β-cyclodextrin (HPBCD). After optimisation of the homogenisation procedure, stable
Zhuomin Zhang et al.
The Analyst, 139(14), 3614-3621 (2014-05-31)
Toxic formaldehyde is sometimes used illegally as a food preservative, however, on-site rapid analysis of trace formaldehyde in aquatic products remains a challenge. In this work, a simple on-site rapid quantification method for trace volatile formaldehyde in aquatic products was

Articles

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service