Skip to Content
Merck
All Photos(1)

Documents

49659

Supelco

2-Methylbutyric acid

analytical standard

Synonym(s):

NSC 7304

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CO2H
CAS Number:
Molecular Weight:
102.13
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.405 (lit.)

bp

176-177 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCC(C)C(O)=O

InChI

1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)

InChI key

WLAMNBDJUVNPJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylbutyric acid belongs to the class of compounds known as organic acids, which are the key components responsible for the quality as well as the nutritional value of food products. It is abundantly found in various fruit-based beverages.

Application

2-Methylbutyric acid may be used as an analytical standard for the determination of the analyte in foodstuffs, wine, and fruit-based beverages by various chromatography techniques.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prevalence, concentration, spoilage, and mitigation of Alicyclobacillus spp. in tropical and subtropical fruit juice concentrates
Danyluk MD, et al.
Food Microbiology, 28(3), 472-477 (2011)
Fast headspace-enantioselective GC-mass spectrometric-multivariate statistical method for routine authentication of flavoured fruit foods
Cagliero C, et al.
Food Chemistry, 132(2), 1071-1079 (2012)
A new method for the determination of short-chain fatty acids from the aliphatic series in wines by headspace solid-phase microextraction-gas chromatography-ion trap mass spectrometry
Olivero SJP and Trujillo JPP
Analytica Chimica Acta, 696(1-2), 59-66 (2011)
Applications of solid-phase microextraction in food analysis
Kataoka H, et al.
Journal of Chromatography A, 880(1-2), 35-62 (2000)
Gas chromatographic determination of 29 organic acids in foodstuffs after continuous solid-phase extraction
Jurado-Sanchez B, et al.
Talanta, 84(3), 924-930 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service