Skip to Content
Merck
All Photos(1)

Documents

19215

Supelco

Butyric acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOH
CAS Number:
Molecular Weight:
88.11
Beilstein:
906770
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.04 (vs air)

vapor pressure

0.43 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

824 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

10 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.398 (lit.)

bp

162 °C (lit.)

mp

−6-−3 °C (lit.)

density

0.964 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCC(O)=O

InChI

1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)

InChI key

FERIUCNNQQJTOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyric acid is a four-carbon containing, saturated short-chain fatty acid. It commonly occurs in its ester form in animal-based fats and plant-based oils. It is a potent differentiation agent for cells and also possesses antiproliferative activity. In cultured mammalian cells, it affects growth rate, morphology, and gene expression. It also induces apoptosis.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

This analytical standard can also be used as follows:

  • Development and validation of a high-performance liquid chromatographic (HPLC) method combined with UV detection to determine four short-chain fatty acids— formic acid, acetic acid, propionic acid, and butyric acid, and lactic acid in bacterial culture samples after their liquid-liquid extraction (LLE)
  • Simultaneous analysis of 10 volatile and non-volatile organic acids in 18 samples of Korean traditional fermented soybean paste, doenjang, by HPLC combined with refractive index detector and gas chromatography coupled with flame ionization detector (GC-FID)
  • Determination of butyric, lactic, acetic, and propionic acids in sour cassava starch waste-water samples by reversed-phase high-performance liquid chromatography (HPLC) and capillary electrophoresis (CE), both coupled with diode array detection (DAD)
  • Multi-residue analysis of six short-chain fatty acids— acetic acid, propionic acid, butyric acid, isobutyric acid, valeric acid, and isovaleric acid, in biological samples of mice colon content by a gas chromatographic method combined with flame ionization detector (GC-FID)
  • Packed-fiber solid phase extraction (PFSPE) of eight short-chain fatty acids using polypyrrole nanofibers, from urine samples of children for quantification by gas chromatography-mass spectrometry (GC-MS)
  • Multi-residue analysis of 76 volatile compounds from jujube extract samples by gas chromatography-mass spectrometry (GC-MS) after their extraction by steam distillation combined with drop-by-drop extraction

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H M Chen et al.
Clinical chemistry, 35(1), 74-76 (1989-01-01)
We describe a procedure for preparing fecal samples for determination of volatile fatty acids (VFAs) by gas-liquid chromatography (GLC) and "high-performance" liquid chromatography (HPLC). The simple, one-step procedure involves only ultrafiltration through a membrane with a molecular-mass cutoff of 3000
Butyric acid: a small fatty acid with diverse biological functions.
K N Prasad
Life sciences, 27(15), 1351-1358 (1980-10-13)
Amita Patnaik et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 8(7), 2142-2148 (2002-07-13)
Pivaloyloxymethyl butyrate (AN-9), an acyloxyalkyl ester prodrug of butyric acid (BA), has demonstrated greater potency than BA at inducing malignant cell differentiation and tumor growth inhibition and has demonstrated more favorable toxicological, pharmacological, and pharmaceutical properties than BA in preclinical
Livia H Morais et al.
Current biology : CB, 30(19), 3761-3774 (2020-08-22)
Birth by Caesarean (C)-section impacts early gut microbiota colonization and is associated with an increased risk of developing immune and metabolic disorders. Moreover, alterations of the microbiome have been shown to affect neurodevelopmental trajectories. However, the long-term effects of C-section
Gili Berkovitch et al.
Journal of medicinal chemistry, 51(23), 7356-7369 (2008-11-15)
Multifunctional acyloxyalkyl ester prodrugs of 5-aminolevulinic acid in cancer cell lines inhibited the proteasome and induced apoptosis and heme synthesis. The most potent prodrug was butyryloxymethyl 5-amino-4-oxopentanoate (1a). The metabolically released formaldehyde from the prodrugs was the dominant factor affecting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service